Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218632103-218632354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLCD4
Entrez Gene ID
84812
Gene Name
phospholipase C delta 4
Gene Aliases
-
UniGene
Hs.632528
Gene Chromosome Location
Chr.2: 218606873-218637186 on Build GRCh38
Transcripts
NM_032726.3 XM_005246913.2 XM_011512012.1 XM_017005114.1 XM_017005115.1 XM_017005116.1 XM_017005117.1 XM_017005118.1 XM_017005119.1 XM_017005120.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5897388 c.1417G>A p.E473K Chr.2: 218632280-218632280 on Build GRCh38
4715049 c.1426C>A p.L476I Chr.2: 218632289-218632289 on Build GRCh38
6414706 c.1440C>A p.D480E Chr.2: 218632303-218632303 on Build GRCh38
3743597 c.1446G>T p.K482N Chr.2: 218632309-218632309 on Build GRCh38
1482768 c.1384G>A p.E462K Chr.2: 218632247-218632247 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770206279 Chr.2: 218632180-218632180 on Build GRCh38
rs763673642 Chr.2: 218632268-218632268 on Build GRCh38
rs752454298 Chr.2: 218632127-218632127 on Build GRCh38
rs777377907 Chr.2: 218632142-218632142 on Build GRCh38
rs369205250 Chr.2: 218632154-218632154 on Build GRCh38
rs761831959 Chr.2: 218632220-218632220 on Build GRCh38
rs776721135 Chr.2: 218632242-218632242 on Build GRCh38
rs61752210 Chr.2: 218632186-218632186 on Build GRCh38
rs377687249 Chr.2: 218632301-218632301 on Build GRCh38
rs145392690 Chr.2: 218632259-218632259 on Build GRCh38
rs373285847 Chr.2: 218632184-218632184 on Build GRCh38
rs747449270 Chr.2: 218632143-218632143 on Build GRCh38
rs749552818 Chr.2: 218632190-218632190 on Build GRCh38
rs370262663 Chr.2: 218632243-218632243 on Build GRCh38
rs746443532 Chr.2: 218632289-218632289 on Build GRCh38
rs781372526 Chr.2: 218632228-218632229 on Build GRCh38
rs780446178 Chr.2: 218632303-218632303 on Build GRCh38
rs748243072 Chr.2: 218632255-218632255 on Build GRCh38
rs3731869 Chr.2: 218632325-218632325 on Build GRCh38
rs774442158 Chr.2: 218632217-218632217 on Build GRCh38
rs375099092 Chr.2: 218632260-218632260 on Build GRCh38
rs953385606 Chr.2: 218632233-218632233 on Build GRCh38
rs757838571 Chr.2: 218632129-218632129 on Build GRCh38
rs539268095 Chr.2: 218632257-218632257 on Build GRCh38
rs751017429 Chr.2: 218632274-218632274 on Build GRCh38
rs746175607 Chr.2: 218632179-218632179 on Build GRCh38
rs756733507 Chr.2: 218632279-218632279 on Build GRCh38
rs766271181 Chr.2: 218632226-218632226 on Build GRCh38
rs557855440 Chr.2: 218632133-218632133 on Build GRCh38
rs749638286 Chr.2: 218632322-218632322 on Build GRCh38
rs765186512 Chr.2: 218632249-218632249 on Build GRCh38
rs758116937 Chr.2: 218632267-218632267 on Build GRCh38
rs997735650 Chr.2: 218632212-218632212 on Build GRCh38
rs781669392 Chr.2: 218632160-218632160 on Build GRCh38
rs554285261 Chr.2: 218632136-218632136 on Build GRCh38
rs768596250 Chr.2: 218632198-218632198 on Build GRCh38
rs189779659 Chr.2: 218632288-218632288 on Build GRCh38

View Full Product Details