Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 34981643-34981906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS1A
Entrez Gene ID
23294
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1A
Gene Aliases
ANKS1
UniGene
Hs.656492
Gene Chromosome Location
Chr.6: 34889261-35097994 on Build GRCh38
Transcripts
NM_015245.2 XM_005248964.3 XM_006715036.2 XM_011514431.2 XM_011514432.2 XM_011514433.1 XM_011514434.2 XM_011514435.2 XM_011514436.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
97996 c.502C>A p.L168I Chr.6: 34981756-34981756 on Build GRCh38
6350746 c.576C>T p.Y192Y Chr.6: 34981830-34981830 on Build GRCh38
5550929 c.464G>T p.C155F Chr.6: 34981718-34981718 on Build GRCh38
4161097 c.482A>T p.H161L Chr.6: 34981736-34981736 on Build GRCh38
5405069 c.603C>T p.L201L Chr.6: 34981857-34981857 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771353885 Chr.6: 34981791-34981791 on Build GRCh38
rs973067545 Chr.6: 34981727-34981727 on Build GRCh38
rs371853881 Chr.6: 34981837-34981837 on Build GRCh38
rs762871214 Chr.6: 34981835-34981835 on Build GRCh38
rs376160942 Chr.6: 34981782-34981782 on Build GRCh38
rs2293242 Chr.6: 34981830-34981830 on Build GRCh38
rs760916630 Chr.6: 34981701-34981701 on Build GRCh38
rs575197928 Chr.6: 34981725-34981725 on Build GRCh38
rs527425948 Chr.6: 34981787-34981787 on Build GRCh38
rs897299237 Chr.6: 34981786-34981786 on Build GRCh38
rs747382147 Chr.6: 34981790-34981790 on Build GRCh38
rs763860633 Chr.6: 34981678-34981678 on Build GRCh38
rs891174948 Chr.6: 34981724-34981724 on Build GRCh38
rs761466250 Chr.6: 34981865-34981865 on Build GRCh38
rs374567948 Chr.6: 34981677-34981677 on Build GRCh38
rs774170415 Chr.6: 34981855-34981855 on Build GRCh38
rs1027642745 Chr.6: 34981827-34981827 on Build GRCh38
rs3817473 Chr.6: 34981674-34981674 on Build GRCh38
rs545384304 Chr.6: 34981773-34981773 on Build GRCh38
rs1037181731 Chr.6: 34981784-34981784 on Build GRCh38
rs779136623 Chr.6: 34981772-34981772 on Build GRCh38
rs755067560 Chr.6: 34981770-34981770 on Build GRCh38
rs769238986 Chr.6: 34981828-34981828 on Build GRCh38
rs200179108 Chr.6: 34981867-34981867 on Build GRCh38
rs753938833 Chr.6: 34981873-34981873 on Build GRCh38
rs371965061 Chr.6: 34981730-34981730 on Build GRCh38
rs752865079 Chr.6: 34981885-34981885 on Build GRCh38
rs914605859 Chr.6: 34981682-34981682 on Build GRCh38
rs143097410 Chr.6: 34981875-34981875 on Build GRCh38
rs932630393 Chr.6: 34981700-34981700 on Build GRCh38
rs942702140 Chr.6: 34981863-34981863 on Build GRCh38
rs766558758 Chr.6: 34981718-34981718 on Build GRCh38
rs936577294 Chr.6: 34981666-34981666 on Build GRCh38
rs367899662 Chr.6: 34981800-34981800 on Build GRCh38
rs776286641 Chr.6: 34981874-34981874 on Build GRCh38
rs769328700 Chr.6: 34981668-34981668 on Build GRCh38
rs751374285 Chr.6: 34981697-34981697 on Build GRCh38
rs745541602 Chr.6: 34981801-34981801 on Build GRCh38

View Full Product Details