Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122833581-122833818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUZD1
Entrez Gene ID
50624
Gene Name
CUB and zona pellucida like domains 1
Gene Aliases
ERG-1, ERG1, ITMAP1, UO-44
UniGene
Hs.647182
Gene Chromosome Location
Chr.10: 122832155-122846175 on Build GRCh38
Transcripts
NM_022034.5 NR_037912.1 NR_037915.1

Gene Symbol
FAM24B-CUZD1
Entrez Gene ID
100533195
Gene Name
FAM24B-CUZD1 readthrough
Gene Aliases
-
UniGene
Hs.647182
Gene Chromosome Location
Chr.10: 122832155-122879641 on Build GRCh38
Transcripts
NM_022034.5 NR_037912.1 NR_037915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
147075 c.1651+5G>T p.? Chr.10: 122833667-122833667 on Build GRCh38
3435334 c.1615C>T p.R539C Chr.10: 122833708-122833708 on Build GRCh38
5012247 c.1565G>A p.R522Q Chr.10: 122833758-122833758 on Build GRCh38
5002869 c.1553C>T p.S518F Chr.10: 122833770-122833770 on Build GRCh38
6567760 c.1531C>A p.R511S Chr.10: 122833792-122833792 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112720477 Chr.10: 122833662-122833662 on Build GRCh38
rs780781010 Chr.10: 122833731-122833731 on Build GRCh38
rs1035427741 Chr.10: 122833688-122833688 on Build GRCh38
rs144406058 Chr.10: 122833698-122833698 on Build GRCh38
rs935378917 Chr.10: 122833760-122833760 on Build GRCh38
rs772196100 Chr.10: 122833692-122833692 on Build GRCh38
rs748037427 Chr.10: 122833636-122833636 on Build GRCh38
rs373802557 Chr.10: 122833792-122833792 on Build GRCh38
rs868328670 Chr.10: 122833758-122833758 on Build GRCh38
rs891718358 Chr.10: 122833652-122833652 on Build GRCh38
rs199609074 Chr.10: 122833745-122833745 on Build GRCh38
rs770986923 Chr.10: 122833781-122833781 on Build GRCh38
rs747160023 Chr.10: 122833778-122833778 on Build GRCh38
rs145074717 Chr.10: 122833686-122833686 on Build GRCh38
rs149964494 Chr.10: 122833678-122833678 on Build GRCh38
rs1011408421 Chr.10: 122833657-122833657 on Build GRCh38
rs556196978 Chr.10: 122833708-122833708 on Build GRCh38
rs969857884 Chr.10: 122833685-122833685 on Build GRCh38
rs773452365 Chr.10: 122833773-122833773 on Build GRCh38
rs925825251 Chr.10: 122833638-122833638 on Build GRCh38
rs142209822 Chr.10: 122833693-122833693 on Build GRCh38
rs752627659 Chr.10: 122833705-122833705 on Build GRCh38
rs755651813 Chr.10: 122833736-122833736 on Build GRCh38
rs372523401 Chr.10: 122833701-122833701 on Build GRCh38
rs142450259 Chr.10: 122833733-122833733 on Build GRCh38
rs375780171 Chr.10: 122833783-122833783 on Build GRCh38
rs772136290 Chr.10: 122833769-122833769 on Build GRCh38
rs376827093 Chr.10: 122833740-122833740 on Build GRCh38
rs540645038 Chr.10: 122833634-122833634 on Build GRCh38
rs749706507 Chr.10: 122833630-122833630 on Build GRCh38
rs749233143 Chr.10: 122833761-122833784 on Build GRCh38
rs368171775 Chr.10: 122833654-122833654 on Build GRCh38
rs1052448647 Chr.10: 122833779-122833779 on Build GRCh38
rs371754237 Chr.10: 122833752-122833752 on Build GRCh38
rs780679975 Chr.10: 122833629-122833629 on Build GRCh38
rs201005450 Chr.10: 122833791-122833791 on Build GRCh38
rs34962077 Chr.10: 122833707-122833707 on Build GRCh38
rs1002086261 Chr.10: 122833713-122833713 on Build GRCh38
rs779078507 Chr.10: 122833755-122833755 on Build GRCh38
rs569019809 Chr.10: 122833759-122833759 on Build GRCh38
rs768113872 Chr.10: 122833784-122833785 on Build GRCh38

View Full Product Details