Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46203712-46203969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR1
Entrez Gene ID
1230
Gene Name
C-C motif chemokine receptor 1
Gene Aliases
CD191, CKR-1, CKR1, CMKBR1, HM145, MIP1aR, SCYAR1
UniGene
Hs.301921
Gene Chromosome Location
Chr.3: 46201709-46208341 on Build GRCh38
Transcripts
NM_001295.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3067993 c.567C>T p.H189H Chr.3: 46203747-46203747 on Build GRCh38
3916119 c.585G>A p.W195* Chr.3: 46203729-46203729 on Build GRCh38
3067998 c.427C>T p.R143W Chr.3: 46203887-46203887 on Build GRCh38
3823961 c.544A>C p.T182P Chr.3: 46203770-46203770 on Build GRCh38
3823962 c.386T>A p.I129N Chr.3: 46203928-46203928 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370179296 Chr.3: 46203746-46203746 on Build GRCh38
rs1027673518 Chr.3: 46203919-46203919 on Build GRCh38
rs754493177 Chr.3: 46203850-46203850 on Build GRCh38
rs776656623 Chr.3: 46203906-46203906 on Build GRCh38
rs758009218 Chr.3: 46203736-46203736 on Build GRCh38
rs768750994 Chr.3: 46203903-46203903 on Build GRCh38
rs200625474 Chr.3: 46203810-46203810 on Build GRCh38
rs772286222 Chr.3: 46203786-46203786 on Build GRCh38
rs201378239 Chr.3: 46203900-46203900 on Build GRCh38
rs762350886 Chr.3: 46203827-46203827 on Build GRCh38
rs779598302 Chr.3: 46203737-46203737 on Build GRCh38
rs200544575 Chr.3: 46203773-46203773 on Build GRCh38
rs765769577 Chr.3: 46203828-46203828 on Build GRCh38
rs3136664 Chr.3: 46203933-46203933 on Build GRCh38
rs199998480 Chr.3: 46203899-46203899 on Build GRCh38
rs1026400808 Chr.3: 46203843-46203843 on Build GRCh38
rs140711017 Chr.3: 46203931-46203931 on Build GRCh38
rs61752589 Chr.3: 46203838-46203838 on Build GRCh38
rs777100916 Chr.3: 46203815-46203815 on Build GRCh38
rs201517958 Chr.3: 46203751-46203751 on Build GRCh38
rs572620512 Chr.3: 46203884-46203884 on Build GRCh38
rs200629341 Chr.3: 46203875-46203875 on Build GRCh38
rs200236244 Chr.3: 46203747-46203747 on Build GRCh38
rs372326161 Chr.3: 46203917-46203917 on Build GRCh38
rs540335334 Chr.3: 46203732-46203732 on Build GRCh38
rs901922371 Chr.3: 46203814-46203814 on Build GRCh38
rs747244913 Chr.3: 46203902-46203902 on Build GRCh38
rs375454466 Chr.3: 46203844-46203844 on Build GRCh38
rs767182036 Chr.3: 46203871-46203871 on Build GRCh38
rs773710336 Chr.3: 46203928-46203928 on Build GRCh38
rs777872590 Chr.3: 46203881-46203881 on Build GRCh38
rs952601094 Chr.3: 46203945-46203945 on Build GRCh38
rs199658220 Chr.3: 46203761-46203761 on Build GRCh38
rs866308207 Chr.3: 46203877-46203877 on Build GRCh38
rs769688894 Chr.3: 46203752-46203752 on Build GRCh38
rs890661710 Chr.3: 46203790-46203790 on Build GRCh38
rs748150820 Chr.3: 46203749-46203749 on Build GRCh38
rs750092631 Chr.3: 46203842-46203842 on Build GRCh38
rs111926601 Chr.3: 46203834-46203834 on Build GRCh38
rs199691125 Chr.3: 46203798-46203798 on Build GRCh38
rs368420597 Chr.3: 46203887-46203887 on Build GRCh38
rs201656427 Chr.3: 46203909-46203909 on Build GRCh38
rs201064013 Chr.3: 46203789-46203789 on Build GRCh38
rs773518362 Chr.3: 46203832-46203832 on Build GRCh38
rs199944458 Chr.3: 46203849-46203849 on Build GRCh38
rs146268408 Chr.3: 46203876-46203876 on Build GRCh38
rs751021323 Chr.3: 46203738-46203738 on Build GRCh38
rs763369579 Chr.3: 46203929-46203929 on Build GRCh38
rs377201514 Chr.3: 46203800-46203800 on Build GRCh38

View Full Product Details