Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 66490433-66490706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP2K1
Entrez Gene ID
5604
Gene Name
mitogen-activated protein kinase kinase 1
Gene Aliases
CFC3, MAPKK1, MEK1, MKK1, PRKMK1
UniGene
Hs.145442
Gene Chromosome Location
Chr.15: 66386873-66491544 on Build GRCh38
Transcripts
NM_002755.3 XM_011521783.2 XM_017022411.1 XM_017022412.1 XM_017022413.1 NM_006049.2

Gene Symbol
SNAPC5
Entrez Gene ID
10302
Gene Name
small nuclear RNA activating complex polypeptide 5
Gene Aliases
SNAP19
UniGene
Hs.30174
Gene Chromosome Location
Chr.15: 66490328-66497808 on Build GRCh38
Transcripts
NM_002755.3 XM_011521783.2 XM_017022411.1 XM_017022412.1 XM_017022413.1 NM_006049.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6785964 c.1069-7C>T p.? Chr.15: 66490495-66490495 on Build GRCh38
1374191 c.1176C>T p.G392G Chr.15: 66490609-66490609 on Build GRCh38
7342441 c.1100A>G p.E367G Chr.15: 66490533-66490533 on Build GRCh38
1214372 c.1180T>C p.*394Q Chr.15: 66490613-66490613 on Build GRCh38
3503334 c.1181A>C p.*394S Chr.15: 66490614-66490614 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751868729 Chr.15: 66490600-66490600 on Build GRCh38
rs540533521 Chr.15: 66490655-66490655 on Build GRCh38
rs371140798 Chr.15: 66490563-66490563 on Build GRCh38
rs767237502 Chr.15: 66490546-66490546 on Build GRCh38
rs781328871 Chr.15: 66490609-66490609 on Build GRCh38
rs574828384 Chr.15: 66490651-66490651 on Build GRCh38
rs376181437 Chr.15: 66490638-66490638 on Build GRCh38
rs534204493 Chr.15: 66490477-66490477 on Build GRCh38
rs755282867 Chr.15: 66490668-66490668 on Build GRCh38
rs781412352 Chr.15: 66490490-66490490 on Build GRCh38
rs759331641 Chr.15: 66490456-66490456 on Build GRCh38
rs752538915 Chr.15: 66490556-66490556 on Build GRCh38
rs28730804 Chr.15: 66490572-66490572 on Build GRCh38
rs759243614 Chr.15: 66490542-66490542 on Build GRCh38
rs200442489 Chr.15: 66490610-66490610 on Build GRCh38
rs558326525 Chr.15: 66490502-66490502 on Build GRCh38
rs752962298 Chr.15: 66490679-66490679 on Build GRCh38
rs748223054 Chr.15: 66490496-66490496 on Build GRCh38
rs751245983 Chr.15: 66490454-66490454 on Build GRCh38
rs201274410 Chr.15: 66490485-66490485 on Build GRCh38
rs761330415 Chr.15: 66490663-66490663 on Build GRCh38
rs763625326 Chr.15: 66490657-66490657 on Build GRCh38
rs771041748 Chr.15: 66490645-66490645 on Build GRCh38
rs776051290 Chr.15: 66490660-66490660 on Build GRCh38
rs770847542 Chr.15: 66490534-66490534 on Build GRCh38
rs755610917 Chr.15: 66490459-66490459 on Build GRCh38
rs767930298 Chr.15: 66490678-66490678 on Build GRCh38
rs896594939 Chr.15: 66490605-66490605 on Build GRCh38
rs769772483 Chr.15: 66490497-66490497 on Build GRCh38
rs766828156 Chr.15: 66490665-66490665 on Build GRCh38
rs750905886 Chr.15: 66490464-66490464 on Build GRCh38
rs537811569 Chr.15: 66490540-66490540 on Build GRCh38
rs568444450 Chr.15: 66490458-66490458 on Build GRCh38
rs372452427 Chr.15: 66490667-66490667 on Build GRCh38
rs756795713 Chr.15: 66490673-66490674 on Build GRCh38
rs941943240 Chr.15: 66490516-66490516 on Build GRCh38
rs756331255 Chr.15: 66490681-66490681 on Build GRCh38
rs1057517863 Chr.15: 66490605-66490605 on Build GRCh38
rs775193561 Chr.15: 66490652-66490652 on Build GRCh38
rs780370635 Chr.15: 66490596-66490596 on Build GRCh38
rs779410802 Chr.15: 66490460-66490460 on Build GRCh38
rs749220939 Chr.15: 66490641-66490641 on Build GRCh38
rs1023819747 Chr.15: 66490639-66490639 on Build GRCh38
rs200293968 Chr.15: 66490531-66490531 on Build GRCh38
rs538726920 Chr.15: 66490495-66490495 on Build GRCh38
rs756208132 Chr.15: 66490614-66490614 on Build GRCh38
rs730880500 Chr.15: 66490601-66490601 on Build GRCh38
rs750934083 Chr.15: 66490571-66490571 on Build GRCh38
rs758866695 Chr.15: 66490576-66490576 on Build GRCh38
rs767106355 Chr.15: 66490457-66490457 on Build GRCh38
rs774452900 Chr.15: 66490646-66490646 on Build GRCh38
rs753352034 Chr.15: 66490474-66490474 on Build GRCh38
rs777783461 Chr.15: 66490632-66490632 on Build GRCh38
rs780460634 Chr.15: 66490478-66490478 on Build GRCh38
rs150841154 Chr.15: 66490570-66490570 on Build GRCh38
rs978417415 Chr.15: 66490672-66490672 on Build GRCh38
rs373794940 Chr.15: 66490568-66490568 on Build GRCh38
rs146942812 Chr.15: 66490486-66490486 on Build GRCh38

View Full Product Details