Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48526922-48527183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBP
Entrez Gene ID
10682
Gene Name
emopamil binding protein (sterol isomerase)
Gene Aliases
CDPX2, CHO2, CPX, CPXD, MEND
UniGene
Hs.30619
Gene Chromosome Location
Chr.X: 48521776-48528716 on Build GRCh38
Transcripts
NM_006579.2

SNP Detail

SNP ID Chromosome Location
rs566252597 Chr.X: 48527121-48527121 on Build GRCh38
rs781809162 Chr.X: 48526973-48526973 on Build GRCh38
rs587783609 Chr.X: 48526997-48526997 on Build GRCh38
rs782274637 Chr.X: 48527052-48527052 on Build GRCh38
rs587783613 Chr.X: 48527015-48527015 on Build GRCh38
rs587783611 Chr.X: 48527001-48527001 on Build GRCh38
rs587783612 Chr.X: 48527007-48527007 on Build GRCh38
rs782325489 Chr.X: 48527056-48527056 on Build GRCh38
rs782118311 Chr.X: 48527073-48527073 on Build GRCh38
rs370841196 Chr.X: 48527069-48527069 on Build GRCh38
rs957046558 Chr.X: 48527070-48527070 on Build GRCh38
rs905580892 Chr.X: 48527023-48527023 on Build GRCh38
rs587783614 Chr.X: 48527018-48527018 on Build GRCh38
rs782635680 Chr.X: 48527064-48527064 on Build GRCh38
rs797045544 Chr.X: 48527019-48527020 on Build GRCh38
rs1010336123 Chr.X: 48527096-48527096 on Build GRCh38
rs782368244 Chr.X: 48527031-48527031 on Build GRCh38
rs587783607 Chr.X: 48526990-48526990 on Build GRCh38
rs587783610 Chr.X: 48526998-48526998 on Build GRCh38
rs781855567 Chr.X: 48526953-48526953 on Build GRCh38
rs1033208894 Chr.X: 48527067-48527067 on Build GRCh38
rs782487869 Chr.X: 48527136-48527136 on Build GRCh38
rs781916312 Chr.X: 48527061-48527061 on Build GRCh38
rs782668236 Chr.X: 48526971-48526971 on Build GRCh38
rs377637290 Chr.X: 48527046-48527046 on Build GRCh38
rs148792902 Chr.X: 48527157-48527157 on Build GRCh38
rs781890344 Chr.X: 48527105-48527105 on Build GRCh38
rs782557120 Chr.X: 48526954-48526954 on Build GRCh38
rs181299601 Chr.X: 48526977-48526977 on Build GRCh38
rs782633217 Chr.X: 48526985-48526985 on Build GRCh38
rs782703576 Chr.X: 48527122-48527122 on Build GRCh38
rs782002953 Chr.X: 48527071-48527071 on Build GRCh38
rs781844882 Chr.X: 48527135-48527135 on Build GRCh38
rs782074263 Chr.X: 48527116-48527116 on Build GRCh38
rs587783608 Chr.X: 48526991-48526991 on Build GRCh38

View Full Product Details