Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37127920-37128193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL2RB
Entrez Gene ID
3560
Gene Name
interleukin 2 receptor subunit beta
Gene Aliases
CD122, IL15RB, P70-75
UniGene
Hs.474787
Gene Chromosome Location
Chr.22: 37125838-37149922 on Build GRCh38
Transcripts
NM_000878.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3554110 c.1608C>T p.Y536Y Chr.22: 37128144-37128144 on Build GRCh38
6685292 c.1636G>T p.D546Y Chr.22: 37128116-37128116 on Build GRCh38
376449 c.1583G>A p.R528H Chr.22: 37128169-37128169 on Build GRCh38
6018242 c.1613C>T p.S538F Chr.22: 37128139-37128139 on Build GRCh38
3972954 c.1629G>T p.Q543H Chr.22: 37128123-37128123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771888990 Chr.22: 37128061-37128061 on Build GRCh38
rs752689706 Chr.22: 37128110-37128110 on Build GRCh38
rs780914414 Chr.22: 37128169-37128169 on Build GRCh38
rs1030806188 Chr.22: 37127960-37127960 on Build GRCh38
rs3218327 Chr.22: 37128004-37128004 on Build GRCh38
rs543241970 Chr.22: 37128055-37128055 on Build GRCh38
rs958049157 Chr.22: 37127988-37127988 on Build GRCh38
rs745463802 Chr.22: 37128051-37128051 on Build GRCh38
rs377234631 Chr.22: 37128170-37128170 on Build GRCh38
rs754271259 Chr.22: 37128094-37128094 on Build GRCh38
rs3218326 Chr.22: 37128052-37128052 on Build GRCh38
rs760940391 Chr.22: 37128077-37128077 on Build GRCh38
rs184918997 Chr.22: 37127953-37127953 on Build GRCh38
rs573251262 Chr.22: 37128113-37128113 on Build GRCh38
rs766780485 Chr.22: 37128084-37128084 on Build GRCh38
rs1030735965 Chr.22: 37127985-37127985 on Build GRCh38
rs751582989 Chr.22: 37128159-37128159 on Build GRCh38
rs777560629 Chr.22: 37128122-37128122 on Build GRCh38
rs968938719 Chr.22: 37127977-37127977 on Build GRCh38
rs376035267 Chr.22: 37128039-37128039 on Build GRCh38
rs969259265 Chr.22: 37128030-37128030 on Build GRCh38
rs758554987 Chr.22: 37128117-37128117 on Build GRCh38
rs774939961 Chr.22: 37128054-37128054 on Build GRCh38
rs759786293 Chr.22: 37128099-37128099 on Build GRCh38
rs538764608 Chr.22: 37128019-37128019 on Build GRCh38
rs1037730914 Chr.22: 37128131-37128131 on Build GRCh38
rs773822329 Chr.22: 37128056-37128056 on Build GRCh38
rs757198038 Chr.22: 37128163-37128163 on Build GRCh38
rs538724059 Chr.22: 37127969-37127969 on Build GRCh38
rs954911321 Chr.22: 37127965-37127965 on Build GRCh38
rs926562990 Chr.22: 37128067-37128067 on Build GRCh38
rs987922301 Chr.22: 37128049-37128049 on Build GRCh38
rs147038391 Chr.22: 37128100-37128100 on Build GRCh38
rs1017813626 Chr.22: 37127970-37127970 on Build GRCh38
rs979100171 Chr.22: 37127982-37127982 on Build GRCh38
rs3218328 Chr.22: 37127968-37127968 on Build GRCh38
rs993580874 Chr.22: 37127950-37127950 on Build GRCh38

View Full Product Details