Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 147981610-147981837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMR1NB
Entrez Gene ID
158521
Gene Name
fragile X mental retardation 1 neighbor
Gene Aliases
CT37, NY-SAR-35, NYSAR35
UniGene
Hs.128580
Gene Chromosome Location
Chr.X: 147981329-148026667 on Build GRCh38
Transcripts
NM_152578.2

SNP Detail

SNP ID Chromosome Location
rs782387330 Chr.X: 147981730-147981730 on Build GRCh38
rs782712428 Chr.X: 147981690-147981690 on Build GRCh38
rs782781690 Chr.X: 147981729-147981729 on Build GRCh38
rs200845775 Chr.X: 147981672-147981672 on Build GRCh38
rs782284194 Chr.X: 147981630-147981630 on Build GRCh38
rs199948782 Chr.X: 147981678-147981678 on Build GRCh38
rs782555748 Chr.X: 147981737-147981737 on Build GRCh38
rs782003415 Chr.X: 147981645-147981645 on Build GRCh38
rs781902910 Chr.X: 147981683-147981683 on Build GRCh38
rs782090366 Chr.X: 147981686-147981686 on Build GRCh38
rs782191396 Chr.X: 147981714-147981714 on Build GRCh38
rs782614686 Chr.X: 147981719-147981719 on Build GRCh38
rs781847857 Chr.X: 147981684-147981684 on Build GRCh38
rs782504719 Chr.X: 147981718-147981718 on Build GRCh38
rs781792716 Chr.X: 147981736-147981736 on Build GRCh38
rs782213986 Chr.X: 147981720-147981720 on Build GRCh38
rs375491250 Chr.X: 147981661-147981661 on Build GRCh38
rs201905210 Chr.X: 147981704-147981704 on Build GRCh38
rs781922096 Chr.X: 147981731-147981732 on Build GRCh38
rs782407330 Chr.X: 147981637-147981637 on Build GRCh38
rs782669082 Chr.X: 147981707-147981707 on Build GRCh38
rs782274311 Chr.X: 147981725-147981725 on Build GRCh38
rs782254799 Chr.X: 147981762-147981762 on Build GRCh38
rs782584943 Chr.X: 147981735-147981735 on Build GRCh38
rs782669957 Chr.X: 147981742-147981742 on Build GRCh38
rs782677931 Chr.X: 147981722-147981722 on Build GRCh38
rs1028827560 Chr.X: 147981763-147981763 on Build GRCh38
rs782245768 Chr.X: 147981646-147981646 on Build GRCh38
rs782225889 Chr.X: 147981725-147981725 on Build GRCh38
rs781842894 Chr.X: 147981731-147981731 on Build GRCh38
rs953298343 Chr.X: 147981773-147981773 on Build GRCh38
rs782335989 Chr.X: 147981719-147981719 on Build GRCh38
rs782314666 Chr.X: 147981721-147981721 on Build GRCh38
rs781942129 Chr.X: 147981670-147981670 on Build GRCh38
rs782434239 Chr.X: 147981673-147981673 on Build GRCh38
rs782487936 Chr.X: 147981734-147981734 on Build GRCh38
rs781869199 Chr.X: 147981716-147981716 on Build GRCh38
rs979405302 Chr.X: 147981787-147981787 on Build GRCh38
rs781786120 Chr.X: 147981699-147981699 on Build GRCh38
rs782135236 Chr.X: 147981679-147981679 on Build GRCh38

View Full Product Details