Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65966147-65966374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SART1
Entrez Gene ID
9092
Gene Name
squamous cell carcinoma antigen recognized by T-cells 1
Gene Aliases
Ara1, HOMS1, SART1259, SNRNP110, Snu66
UniGene
Hs.502883
Gene Chromosome Location
Chr.11: 65961689-65980136 on Build GRCh38
Transcripts
NM_005146.4 XM_011545344.1 XM_011545345.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7294857 c.981+2C>T p.? Chr.11: 65966220-65966220 on Build GRCh38
6771957 c.981+4C>T p.? Chr.11: 65966222-65966222 on Build GRCh38
6134235 c.992G>T p.R331L Chr.11: 65966360-65966360 on Build GRCh38
378972 c.964G>A p.V322M Chr.11: 65966201-65966201 on Build GRCh38
930592 c.990T>C p.P330P Chr.11: 65966358-65966358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200457088 Chr.11: 65966343-65966343 on Build GRCh38
rs774370584 Chr.11: 65966284-65966284 on Build GRCh38
rs770994883 Chr.11: 65966328-65966328 on Build GRCh38
rs774591944 Chr.11: 65966324-65966324 on Build GRCh38
rs757816096 Chr.11: 65966248-65966248 on Build GRCh38
rs776510670 Chr.11: 65966336-65966336 on Build GRCh38
rs763789733 Chr.11: 65966266-65966266 on Build GRCh38
rs768843222 Chr.11: 65966197-65966197 on Build GRCh38
rs111521324 Chr.11: 65966214-65966214 on Build GRCh38
rs752014238 Chr.11: 65966266-65966266 on Build GRCh38
rs747555166 Chr.11: 65966168-65966168 on Build GRCh38
rs763711048 Chr.11: 65966259-65966259 on Build GRCh38
rs542303302 Chr.11: 65966244-65966244 on Build GRCh38
rs202095327 Chr.11: 65966206-65966206 on Build GRCh38
rs923170569 Chr.11: 65966201-65966201 on Build GRCh38
rs781559975 Chr.11: 65966180-65966180 on Build GRCh38
rs768594116 Chr.11: 65966317-65966317 on Build GRCh38
rs778139903 Chr.11: 65966165-65966165 on Build GRCh38
rs369954244 Chr.11: 65966251-65966251 on Build GRCh38
rs201006690 Chr.11: 65966200-65966200 on Build GRCh38
rs781471797 Chr.11: 65966279-65966279 on Build GRCh38
rs765122173 Chr.11: 65966341-65966341 on Build GRCh38
rs561904217 Chr.11: 65966304-65966304 on Build GRCh38
rs751126511 Chr.11: 65966313-65966313 on Build GRCh38
rs141499780 Chr.11: 65966215-65966215 on Build GRCh38
rs757796506 Chr.11: 65966272-65966272 on Build GRCh38
rs1013732740 Chr.11: 65966312-65966312 on Build GRCh38
rs775586538 Chr.11: 65966194-65966194 on Build GRCh38
rs751077055 Chr.11: 65966347-65966347 on Build GRCh38
rs147105474 Chr.11: 65966188-65966188 on Build GRCh38
rs887644299 Chr.11: 65966262-65966262 on Build GRCh38
rs762725337 Chr.11: 65966244-65966244 on Build GRCh38
rs200289737 Chr.11: 65966220-65966220 on Build GRCh38
rs530721949 Chr.11: 65966338-65966338 on Build GRCh38
rs771265194 Chr.11: 65966179-65966179 on Build GRCh38
rs764810513 Chr.11: 65966236-65966236 on Build GRCh38
rs201529113 Chr.11: 65966342-65966342 on Build GRCh38
rs759197059 Chr.11: 65966222-65966222 on Build GRCh38
rs141881804 Chr.11: 65966186-65966186 on Build GRCh38
rs746902054 Chr.11: 65966325-65966325 on Build GRCh38
rs749561285 Chr.11: 65966308-65966308 on Build GRCh38
rs756563602 Chr.11: 65966301-65966301 on Build GRCh38
rs201368685 Chr.11: 65966189-65966189 on Build GRCh38

View Full Product Details