Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 116854342-116854581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD21
Entrez Gene ID
5885
Gene Name
RAD21 cohesin complex component
Gene Aliases
CDLS4, HR21, HRAD21, MCD1, NXP1, SCC1, hHR21
UniGene
Hs.81848
Gene Chromosome Location
Chr.8: 116845934-116874866 on Build GRCh38
Transcripts
NM_006265.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3768667 c.1064C>T p.P355L Chr.8: 116854342-116854342 on Build GRCh38
5879120 c.990_991insA p.E331fs*4 Chr.8: 116854415-116854416 on Build GRCh38
3778851 c.1005G>C p.K335N Chr.8: 116854401-116854401 on Build GRCh38
6946233 c.943G>T p.E315* Chr.8: 116854463-116854463 on Build GRCh38
5944547 c.972_973insG p.I325fs*3 Chr.8: 116854433-116854434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200333487 Chr.8: 116854479-116854479 on Build GRCh38
rs201146181 Chr.8: 116854392-116854392 on Build GRCh38
rs765764891 Chr.8: 116854511-116854511 on Build GRCh38
rs767878994 Chr.8: 116854519-116854520 on Build GRCh38
rs375637600 Chr.8: 116854491-116854491 on Build GRCh38
rs10101469 Chr.8: 116854541-116854541 on Build GRCh38
rs893432919 Chr.8: 116854499-116854499 on Build GRCh38
rs778800442 Chr.8: 116854437-116854437 on Build GRCh38
rs377667234 Chr.8: 116854509-116854509 on Build GRCh38
rs774690551 Chr.8: 116854482-116854482 on Build GRCh38
rs768725843 Chr.8: 116854407-116854407 on Build GRCh38
rs938268975 Chr.8: 116854420-116854420 on Build GRCh38
rs917737700 Chr.8: 116854556-116854556 on Build GRCh38
rs748245537 Chr.8: 116854465-116854465 on Build GRCh38
rs761168258 Chr.8: 116854486-116854486 on Build GRCh38
rs267601737 Chr.8: 116854372-116854372 on Build GRCh38
rs908977328 Chr.8: 116854514-116854514 on Build GRCh38
rs771370772 Chr.8: 116854490-116854490 on Build GRCh38
rs902290392 Chr.8: 116854548-116854548 on Build GRCh38
rs761355472 Chr.8: 116854515-116854515 on Build GRCh38
rs772246627 Chr.8: 116854477-116854477 on Build GRCh38
rs762219753 Chr.8: 116854495-116854495 on Build GRCh38

View Full Product Details