Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 230200810-230201059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SP110
Entrez Gene ID
3431
Gene Name
SP110 nuclear body protein
Gene Aliases
IFI41, IFI75, IPR1, VODI
UniGene
Hs.145150
Gene Chromosome Location
Chr.2: 230165466-230225729 on Build GRCh38
Transcripts
NM_001185015.1 NM_004509.3 NM_004510.3 NM_080424.2 XM_005246525.3 XM_006712487.3 XM_006712489.3 XM_011511088.2 XM_011511089.2 XM_011511090.2 XM_011511091.2 XM_011511092.2 XM_017003968.1 XM_017003969.1 XM_017003970.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7423180 c.1139T>C p.V380A Chr.2: 230200893-230200893 on Build GRCh38
7124177 c.1067C>T p.S356L Chr.2: 230200947-230200947 on Build GRCh38
3048255 c.1092G>A p.M364I Chr.2: 230200940-230200940 on Build GRCh38
7423179 c.1121T>C p.V374A Chr.2: 230200893-230200893 on Build GRCh38
7391495 c.1118C>T p.T373M Chr.2: 230200914-230200914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770884627 Chr.2: 230200934-230200934 on Build GRCh38
rs201182994 Chr.2: 230200868-230200868 on Build GRCh38
rs200571911 Chr.2: 230201019-230201020 on Build GRCh38
rs569159319 Chr.2: 230200884-230200884 on Build GRCh38
rs773736979 Chr.2: 230200998-230200998 on Build GRCh38
rs369764018 Chr.2: 230200919-230200919 on Build GRCh38
rs758333221 Chr.2: 230200874-230200874 on Build GRCh38
rs201350600 Chr.2: 230200905-230200905 on Build GRCh38
rs764054594 Chr.2: 230200881-230200881 on Build GRCh38
rs377705679 Chr.2: 230201001-230201001 on Build GRCh38
rs770087375 Chr.2: 230200877-230200877 on Build GRCh38
rs775810248 Chr.2: 230200947-230200947 on Build GRCh38
rs1008864778 Chr.2: 230200925-230200925 on Build GRCh38
rs41309108 Chr.2: 230201006-230201006 on Build GRCh38
rs774279478 Chr.2: 230200970-230200970 on Build GRCh38
rs200621430 Chr.2: 230200913-230200913 on Build GRCh38
rs754241391 Chr.2: 230201007-230201007 on Build GRCh38
rs199713399 Chr.2: 230200900-230200900 on Build GRCh38
rs778860082 Chr.2: 230200854-230200854 on Build GRCh38
rs200460698 Chr.2: 230200843-230200843 on Build GRCh38
rs567078797 Chr.2: 230201021-230201021 on Build GRCh38
rs760607836 Chr.2: 230200876-230200876 on Build GRCh38
rs539213557 Chr.2: 230200981-230200981 on Build GRCh38
rs777870124 Chr.2: 230200922-230200922 on Build GRCh38
rs201394862 Chr.2: 230200899-230200899 on Build GRCh38
rs373007586 Chr.2: 230200946-230200946 on Build GRCh38
rs199526783 Chr.2: 230200973-230200973 on Build GRCh38
rs1014974380 Chr.2: 230200960-230200960 on Build GRCh38
rs764999776 Chr.2: 230201017-230201017 on Build GRCh38
rs768985896 Chr.2: 230200959-230200959 on Build GRCh38
rs762218275 Chr.2: 230200887-230200887 on Build GRCh38
rs962546053 Chr.2: 230200912-230200912 on Build GRCh38
rs59573011 Chr.2: 230200914-230200914 on Build GRCh38
rs748015488 Chr.2: 230200858-230200858 on Build GRCh38
rs1030840458 Chr.2: 230200955-230200955 on Build GRCh38
rs754583325 Chr.2: 230201010-230201010 on Build GRCh38
rs749437023 Chr.2: 230200958-230200958 on Build GRCh38
rs201953042 Chr.2: 230200869-230200869 on Build GRCh38
rs200263454 Chr.2: 230200893-230200893 on Build GRCh38
rs775861638 Chr.2: 230200878-230200878 on Build GRCh38
rs550857501 Chr.2: 230200841-230200841 on Build GRCh38
rs1027472092 Chr.2: 230201009-230201009 on Build GRCh38
rs199620617 Chr.2: 230200909-230200909 on Build GRCh38
rs768123625 Chr.2: 230200888-230200888 on Build GRCh38
rs746736624 Chr.2: 230200923-230200923 on Build GRCh38
rs200627866 Chr.2: 230200903-230200903 on Build GRCh38
rs768081054 Chr.2: 230200851-230200851 on Build GRCh38

View Full Product Details