Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130614195-130614444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUBP3
Entrez Gene ID
8939
Gene Name
far upstream element binding protein 3
Gene Aliases
FBP3
UniGene
Hs.673029
Gene Chromosome Location
Chr.9: 130579567-130638352 on Build GRCh38
Transcripts
NM_003934.1 XM_005272232.1 XM_006717313.2 XM_011519172.2 XM_011519173.2 XM_011519174.2 XM_017015269.1 XM_017015270.1 XM_017015271.1 XM_017015272.1 XM_017015273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5467626 c.224+4A>C p.? Chr.9: 130614349-130614349 on Build GRCh38
7309124 c.224+5G>C p.? Chr.9: 130614350-130614350 on Build GRCh38
6182784 c.188G>A p.R63K Chr.9: 130614309-130614309 on Build GRCh38
6378204 c.224A>T p.E75V Chr.9: 130614345-130614345 on Build GRCh38
396334 c.391C>T p.P131S Chr.9: 130614332-130614332 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956878159 Chr.9: 130614234-130614234 on Build GRCh38
rs771614602 Chr.9: 130614333-130614333 on Build GRCh38
rs1042549900 Chr.9: 130614241-130614242 on Build GRCh38
rs767576730 Chr.9: 130614268-130614268 on Build GRCh38
rs942394677 Chr.9: 130614325-130614325 on Build GRCh38
rs771363680 Chr.9: 130614256-130614256 on Build GRCh38
rs924112460 Chr.9: 130614296-130614296 on Build GRCh38
rs747070899 Chr.9: 130614248-130614248 on Build GRCh38
rs577946962 Chr.9: 130614373-130614373 on Build GRCh38
rs781682860 Chr.9: 130614265-130614265 on Build GRCh38
rs758598148 Chr.9: 130614245-130614245 on Build GRCh38
rs765100555 Chr.9: 130614389-130614389 on Build GRCh38
rs763860061 Chr.9: 130614267-130614269 on Build GRCh38
rs746225075 Chr.9: 130614266-130614266 on Build GRCh38
rs748365429 Chr.9: 130614242-130614242 on Build GRCh38
rs753006097 Chr.9: 130614396-130614396 on Build GRCh38
rs758724707 Chr.9: 130614397-130614397 on Build GRCh38
rs1051352129 Chr.9: 130614326-130614326 on Build GRCh38
rs779342895 Chr.9: 130614238-130614238 on Build GRCh38
rs760169894 Chr.9: 130614363-130614363 on Build GRCh38
rs759304186 Chr.9: 130614387-130614387 on Build GRCh38
rs761435847 Chr.9: 130614324-130614324 on Build GRCh38
rs770140899 Chr.9: 130614280-130614280 on Build GRCh38
rs772743849 Chr.9: 130614353-130614353 on Build GRCh38
rs753807291 Chr.9: 130614370-130614370 on Build GRCh38
rs749886402 Chr.9: 130614365-130614365 on Build GRCh38
rs538792338 Chr.9: 130614246-130614246 on Build GRCh38
rs774051133 Chr.9: 130614323-130614323 on Build GRCh38

View Full Product Details