Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 20159783-20160016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBX1
Entrez Gene ID
120237
Gene Name
developing brain homeobox 1
Gene Aliases
-
UniGene
Hs.558604
Gene Chromosome Location
Chr.11: 20156214-20160324 on Build GRCh38
Transcripts
NM_001029865.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3979498 c.367+2T>G p.? Chr.11: 20159956-20159956 on Build GRCh38
232131 c.350C>T p.S117F Chr.11: 20159975-20159975 on Build GRCh38
1297922 c.351C>T p.S117S Chr.11: 20159974-20159974 on Build GRCh38
5951114 c.367+6C>T p.? Chr.11: 20159952-20159952 on Build GRCh38
74326 c.364A>G p.T122A Chr.11: 20159961-20159961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769483173 Chr.11: 20159930-20159930 on Build GRCh38
rs545095950 Chr.11: 20159864-20159864 on Build GRCh38
rs1014771799 Chr.11: 20159818-20159818 on Build GRCh38
rs781301260 Chr.11: 20159920-20159920 on Build GRCh38
rs775978418 Chr.11: 20159969-20159969 on Build GRCh38
rs754071944 Chr.11: 20159977-20159977 on Build GRCh38
rs950995954 Chr.11: 20159891-20159892 on Build GRCh38
rs762561545 Chr.11: 20159936-20159936 on Build GRCh38
rs565091187 Chr.11: 20159866-20159866 on Build GRCh38
rs767950740 Chr.11: 20159968-20159968 on Build GRCh38
rs765531264 Chr.11: 20159986-20159986 on Build GRCh38
rs1006442150 Chr.11: 20159907-20159907 on Build GRCh38
rs374624813 Chr.11: 20159948-20159948 on Build GRCh38
rs757464120 Chr.11: 20159985-20159985 on Build GRCh38
rs193082081 Chr.11: 20159836-20159836 on Build GRCh38
rs768674515 Chr.11: 20159909-20159909 on Build GRCh38
rs367554777 Chr.11: 20159949-20159949 on Build GRCh38
rs781018569 Chr.11: 20159933-20159933 on Build GRCh38
rs185823620 Chr.11: 20159879-20159879 on Build GRCh38
rs750481336 Chr.11: 20159989-20159989 on Build GRCh38
rs868480621 Chr.11: 20159975-20159975 on Build GRCh38
rs200102447 Chr.11: 20159929-20159929 on Build GRCh38
rs746916471 Chr.11: 20159908-20159908 on Build GRCh38
rs747895080 Chr.11: 20159922-20159922 on Build GRCh38
rs371391865 Chr.11: 20159954-20159954 on Build GRCh38
rs907059036 Chr.11: 20159826-20159826 on Build GRCh38
rs879393652 Chr.11: 20159875-20159875 on Build GRCh38
rs772904148 Chr.11: 20159935-20159935 on Build GRCh38
rs764426810 Chr.11: 20159972-20159972 on Build GRCh38
rs1004805355 Chr.11: 20159806-20159806 on Build GRCh38
rs959525364 Chr.11: 20159901-20159901 on Build GRCh38
rs761019823 Chr.11: 20159971-20159971 on Build GRCh38

View Full Product Details