Product References
Oocyte Meiotic Competence in the Domestic Cat Model: Novel Roles for Nuclear Proteins BRD2 and NPM1.
Frontiers in cell and developmental biology
Chavez DR,Lee PC,Comizzoli P
Published figure using beta Tubulin monoclonal antibody (Product # MA5-11732) in Immunocytochemistry
Tue Jun 08 00:00:00 EDT 2021
Oocyte Meiotic Competence in the Domestic Cat Model: Novel Roles for Nuclear Proteins BRD2 and NPM1.
Frontiers in cell and developmental biology
Chavez DR,Lee PC,Comizzoli P
Published figure using beta Tubulin monoclonal antibody (Product # MA5-11732) in Immunocytochemistry
Tue Jun 08 00:00:00 EDT 2021
Oocyte Meiotic Competence in the Domestic Cat Model: Novel Roles for Nuclear Proteins BRD2 and NPM1.
Frontiers in cell and developmental biology
Chavez DR,Lee PC,Comizzoli P
Published figure using beta Tubulin monoclonal antibody (Product # MA5-11732) in Immunocytochemistry
Tue Jun 08 00:00:00 EDT 2021
Ganoderic Acid A Attenuates LPS-Induced Neuroinflammation in BV2 Microglia by Activating Farnesoid X Receptor.
Neurochemical research
Jia Y,Zhang D,Yin H,Li H,Du J,Bao H
MA5-11732 was used in Western Blot to demonstrate that GAA can significantly inhibit LPS-induced neuroinflammation in BV2 microglial cells via activating FXR receptor.
Thu Jul 01 00:00:00 EDT 2021
Genistein-3'-sodium sulfonate Attenuates Neuroinflammation in Stroke Rats by Down-Regulating Microglial M1 Polarization through α7nAChR-NF-κB Signaling Pathway.
International journal of biological sciences
Liu C,Liu S,Xiong L,Zhang L,Li X,Cao X,Xue J,Li L,Huang C,Huang Z
Published figure using beta Tubulin monoclonal antibody (Product # MA5-11732) in Western Blot
Fri Jan 21 00:00:00 EST 2022
Antioxidant Strategy to Prevent Simulated Microgravity-Induced Effects on Bone Osteoblasts.
International journal of molecular sciences
Morabito C,Guarnieri S,Cucina A,Bizzarri M,Mariggiò MA
Published figure using beta Tubulin monoclonal antibody (Product # MA5-11732) in Western Blot
Thu May 21 00:00:00 EDT 2020
Antioxidant Strategy to Prevent Simulated Microgravity-Induced Effects on Bone Osteoblasts.
International journal of molecular sciences
Morabito C,Guarnieri S,Cucina A,Bizzarri M,Mariggiò MA
Published figure using beta Tubulin monoclonal antibody (Product # MA5-11732) in Western Blot
Thu May 21 00:00:00 EDT 2020
Ikzf1 regulates embryonic T lymphopoiesis via Ccr9 and Irf4 in zebrafish.
The Journal of biological chemistry
Huang Y,Lu Y,He Y,Feng Z,Zhan Y,Huang X,Liu Q,Zhang J,Li H,Huang H,Ma M,Luo L,Li L
MA5-11732 was used in Western Blotting to report that the genetic ablation of ikzf1 in mutant zebrafish resulted in abrogated embryonic T lymphopoiesis.
Fri Nov 01 00:00:00 EDT 2019
Human ApoE Isoforms Differentially Modulate Brain Glucose and Ketone Body Metabolism: Implications for Alzheimer's Disease Risk Reduction and Early Intervention.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Wu L,Zhang X,Zhao L
MA5-11732 was used in Western Blotting to investigate the impact of human apolipoprotein E isoforms on brain energy metabolism, an area significantly perturbed in preclinical Alzheimer's disease.
Wed Jul 25 00:00:00 EDT 2018
FoxM1 drives ADAM17/EGFR activation loop to promote mesenchymal transition in glioblastoma.
Cell death & disease
Zhang C,Han X,Xu X,Zhou Z,Chen X,Tang Y,Cheng J,Moazzam NF,Liu F,Xu J,Peng W,Du F,Zhang B,Song Z,Zeng J,Gong A
Published figure using beta Tubulin monoclonal antibody (Product # MA5-11732) in Western Blot
Tue May 01 00:00:00 EDT 2018
Influence of warming and reanimation conditions on seminiferous tubule morphology, mitochondrial activity, and cell composition of vitrified testicular tissues in the domestic cat model.
PloS one
Lima DBC,Silva LDMD,Comizzoli P
MA5-11732 was used in Western Blotting to study structural and functional properties of testicular tissues from prepubertal domestic cats after standard vitrification followed by two warming protocols.
Tue Apr 16 00:00:00 EDT 2019
A bacterial type III secretion-based protein delivery tool for broad applications in cell biology.
The Journal of cell biology
Ittig SJ,Schmutz C,Kasper CA,Amstutz M,Schmidt A,Sauteur L,Vigano MA,Low SH,Affolter M,Cornelis GR,Nigg EA,Arrieumerlou C
Published figure using beta Tubulin monoclonal antibody (Product # MA5-11732) in Immunofluorescence
Mon Nov 23 00:00:00 EST 2015
Live imaging of prions reveals nascent PrPSc in cell-surface, raft-associated amyloid strings and webs.
The Journal of cell biology
Rouvinski A,Karniely S,Kounin M,Moussa S,Goldberg MD,Warburg G,Lyakhovetsky R,Papy-Garcia D,Kutzsche J,Korth C,Carlson GA,Godsave SF,Peters PJ,Luhr K,Kristensson K,Taraboulos A
MA5-11732 was used in immunocytochemistry to study the existence of PrPSC in cell surface strings and webs associated with lipid rafts using live imaging methodologies
Mon Feb 03 00:00:00 EST 2014
Human subtilase SKI-1/S1P is a master regulator of the HCV Lifecycle and a potential host cell target for developing indirect-acting antiviral agents.
PLoS pathogens
Olmstead AD,Knecht W,Lazarov I,Dixit SB,Jean F
MA5-11732 was used in western blot to study the role of human subtilase SKI-1/S1P in the hepatitis C virus lifecycle
Sun Jan 01 00:00:00 EST 2012
5-Aminoimidazole-4-carboxyamide ribonucleoside induces G(1)/S arrest and Nanog downregulation via p53 and enhances erythroid differentiation.
Stem cells (Dayton, Ohio)
Chae HD,Lee MR,Broxmeyer HE
MA5-11732 was used in western blot to study the role of p53 in the mechanism by which the AMPK activator AICAR induces cell cycle arrest and promotes erythroid differentiation in murine embryonic stem cells
Wed Feb 01 00:00:00 EST 2012
Sphingosylphosphorylcholine induces stress fiber formation via activation of Fyn-RhoA-ROCK signaling pathway in fibroblasts.
Cellular signalling
Xu D,Kishi H,Kawamichi H,Kajiya K,Takada Y,Kobayashi S
MA5-11732 was used in western blot to investigate the involvement of Fyn-RhoA-ROCK signaling pathway in the effect of sphingosylphosphorylcholine on stress fiber formation
Sun Jan 01 00:00:00 EST 2012
Mechanism-based screen for G1/S checkpoint activators identifies a selective activator of EIF2AK3/PERK signalling.
PloS one
Stockwell SR,Platt G,Barrie SE,Zoumpoulidou G,Te Poele RH,Aherne GW,Wilson SC,Sheldrake P,McDonald E,Venet M,Soudy C,Elustondo F,Rigoreau L,Blagg J,Workman P,Garrett MD,Mittnacht S
MA5-11732 was used in western blot to identify and characterize small molecule inhibitors of G1/S checkpoint activation
Tue May 29 00:00:00 EDT 2012
Cytosolic carboxypeptidase 1 is involved in processing α- and β-tubulin.
The Journal of biological chemistry
Berezniuk I,Vu HT,Lyons PJ,Sironi JJ,Xiao H,Burd B,Setou M,Angeletti RH,Ikegami K,Fricker LD
MA5-11732 was used in western blot to study the role of cytosolic carboxypeptidase 1 in processing Glu residues in ?- and ?-tubulin
Fri Feb 24 00:00:00 EST 2012
Severe osteogenesis imperfecta caused by a small in-frame deletion in CRTAP.
American journal of medical genetics. Part A
Amor IM,Rauch F,Gruenwald K,Weis M,Eyre DR,Roughley P,Glorieux FH,Morello R
MA5-11732 was used in Western Blotting to characterise a patient with a cartilage-associated protein small in-frame deletion, causing severe osteogenesis imperfecta.
Tue Nov 01 00:00:00 EDT 2011
Trastuzumab induced in vivo tissue remodelling associated in vitro with inhibition of the active forms of AKT and PTEN and RhoB induction in an ovarian carcinoma model.
British journal of cancer
Delord JP,Quideau S,Rochaix P,Caselles O,Couderc B,Hennebelle I,Courbon F,Canal P,Allal BC
MA5-11732 was used in western blot to study the biological, molecular and cellular effects of Herceptin in ovarian carcinoma
Tue Jun 29 00:00:00 EDT 2010
HIV-1 Vpr induces TLR4/MyD88-mediated IL-6 production and reactivates viral production from latency.
Journal of leukocyte biology
Hoshino S,Konishi M,Mori M,Shimura M,Nishitani C,Kuroki Y,Koyanagi Y,Kano S,Itabe H,Ishizaka Y
MA5-11732 was used in western blot to investigate the effect of HIV-1 Vpr on interleukin 6 production and viral production
Tue Jun 01 00:00:00 EDT 2010
A novel human dynactin-associated protein, dynAP, promotes activation of Akt, and ergosterol-related compounds induce dynAP-dependent apoptosis of human cancer cells.
Molecular cancer therapeutics
Kunoh T,Noda T,Koseki K,Sekigawa M,Takagi M,Shin-ya K,Goshima N,Iemura S,Natsume T,Wada S,Mukai Y,Ohta S,Sasaki R,Mizukami T
MA5-11732 was used in immunocytochemistry to study the role dynAP, a dynactin-associated protein, in the induction of human cancer cell apoptosis by ergosterol-related compounds
Mon Nov 01 00:00:00 EDT 2010
L-type calcium channel blockers reverse docetaxel and vincristine-induced multidrug resistance independent of ABCB1 expression in human lung cancer cell lines.
Toxicology letters
Chiu LY,Ko JL,Lee YJ,Yang TY,Tee YT,Sheu GT
MA5-11732 was used in western blot to investigate the relationship between ABCB1 expression and multidrug resistance induced by docetaxel or vincristine in human lung cancer cell lines
Mon Feb 15 00:00:00 EST 2010
Modulation of TNF-alpha-converting enzyme by the spike protein of SARS-CoV and ACE2 induces TNF-alpha production and facilitates viral entry.
Proceedings of the National Academy of Sciences of the United States of America
Haga S,Yamamoto N,Nakai-Murakami C,Osawa Y,Tokunaga K,Sata T,Yamamoto N,Sasazuki T,Ishizaka Y
MA5-11732 was used in western blot to the role of SARS-CoV spike protein in the downregulation of angiotensin-converting enzyme 2 and its mechanism
Tue Jun 03 00:00:00 EDT 2008
KIT overexpression induces proliferation in astrocytes in an imatinib-responsive manner and associates with proliferation index in gliomas.
International journal of cancer
Blom T,Fox H,Angers-Loustau A,Peltonen K,Kerosuo L,Wartiovaara K,Linja M,Jänne OA,Kovanen P,Haapasalo H,Nupponen NN
MA5-11732 was used in western blot to investigate the mechanism for the induction of astrocyte proliferation by KIT
Fri Aug 15 00:00:00 EDT 2008
Dynamic regulation of endothelial NOS mediated by competitive interaction with alpha-actinin-4 and calmodulin.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Hiroi Y,Guo Z,Li Y,Beggs AH,Liao JK
MA5-11732 was used in western blot to investigate the mechanism of eNOS activity regulation in vascular endothelial cells
Thu May 01 00:00:00 EDT 2008
Progressive changes in hepatoma cells stably transfected with hepatitis B virus X gene.
Intervirology
Ye L,Dong N,Wang Q,Xu Z,Cai N,Wang H,Zhang X
MA5-11732 was used in western blot to study the molecular mechanism of hepatocellular carcinoma development induced by hepatitis B virus X protein
Fri Apr 18 00:00:00 EDT 2008
Characterization of t(6;11)(p21;q12) in a renal-cell carcinoma of an adult patient.
Genes, chromosomes & cancer
Pecciarini L,Cangi MG,Lo Cunsolo C,Macri' E,Dal Cin E,Martignoni G,Doglioni C
MA5-11732 was used in western blot to report a clinical case of t(6;11)(p21;q12) renal-cell carcinoma
Tue May 01 00:00:00 EDT 2007
High expression of integrin beta1 correlates with high proliferation capacity in oral keratinocytes.
Wiener klinische Wochenschrift
Stein E,Blaimauer K,Bauer S,Erovic BM,Turhani D,Thurnher D
MA5-11732 was used in western blot to isolate two fractions of oral keratinocytes based on their affinity to collagen type IV to obtain cells with high prolifrative potential
Mon Nov 05 00:00:00 EST 2007
S100A7 (Psoriasin), highly expressed in ductal carcinoma in situ (DCIS), is regulated by IFN-gamma in mammary epithelial cells.
BMC cancer
Petersson S,Bylander A,Yhr M,Enerbäck C
MA5-11732 was used in western blot to study the signal transduction pathways involved in the regulation of psoriasin in murine mammary epithelial cells
Tue Nov 06 00:00:00 EST 2007
The topoisomerase II-Hsp90 complex: a new chemotherapeutic target?
International journal of cancer
Barker CR,Hamlett J,Pennington SR,Burrows F,Lundgren K,Lough R,Watson AJ,Jenkins JR
MA5-11732 was used in immunoprecipitation and western blot to study the topoisomerase II-Hsp90 complex as a potential new chemotherapeutic target
Thu Jun 01 00:00:00 EDT 2006
CDK-dependent activation of poly(ADP-ribose) polymerase member 10 (PARP10).
The Journal of biological chemistry
Chou HY,Chou HT,Lee SC
MA5-11732 was used in western blot to study the CDK-dependent activation of poly(ADP-ribose) polymerase member 10 (PARP10)
Fri Jun 02 00:00:00 EDT 2006
Upregulation of tissue inhibitor of metalloproteinases (TIMP)-2 promotes matrix metalloproteinase (MMP)-2 activation and cell invasion in a human glioblastoma cell line.
Laboratory investigation; a journal of technical methods and pathology
Lu KV,Jong KA,Rajasekaran AK,Cloughesy TF,Mischel PS
MA5-11732 was used in western blot to study the role of TIMP-2 upregulation in promoting MMP2 activation and cell invasion in a human glioblastoma cell line
Thu Jan 01 00:00:00 EST 2004
Differential segregation and modification of mRNA during spermiogenesis in Marsilea vestita.
Developmental biology
Tsai CW,Van Der Weele CM,Wolniak SM
MA5-11732 was used in western blot to study the differential segregation and modification of mRNA during spermiogenesis in Marsilea vestita
Sat May 15 00:00:00 EDT 2004
Altered levels and regulation of stathmin in paclitaxel-resistant ovarian cancer cells.
Oncogene
Balachandran R,Welsh MJ,Day BW
MA5-11732 was used in western blot to investigate the changes of stathmin in specific cancer cell lines
Thu Dec 04 00:00:00 EST 2003