Product References
Complement-activating IgM enhances the humoral but not the T cell immune response in mice.
PloS one
Ding Z,Bergman A,Rutemark C,Ouchida R,Ohno H,Wang JY,Heyman B
MM7501 was used in flow cytometry to confirm the importance of complement for the ability of specific IgM to enhance antibody responses.
Mon Mar 30 00:00:00 EDT 2015
Immunosuppressive drugs on inducing Ag-specific CD4(+)CD25(+)Foxp3(+) Treg cells during immune response in vivo.
Transplant immunology
Wu T,Zhang L,Xu K,Sun C,Lei T,Peng J,Liu G,Wang R,Zhao Y
MM7501 was used in flow cytometry to examine the impact of immunosuppressants on Ag-specific Tregs during immune response.
Wed Aug 01 00:00:00 EDT 2012
Graded attenuation of TCR signaling elicits distinct autoimmune diseases by altering thymic T cell selection and regulatory T cell function.
Journal of immunology (Baltimore, Md. : 1950)
Tanaka S,Maeda S,Hashimoto M,Fujimori C,Ito Y,Teradaira S,Hirota K,Yoshitomi H,Katakai T,Shimizu A,Nomura T,Sakaguchi N,Sakaguchi S
MM7501 was used in flow cytometry to characterize T cells and arthritis using +/+, skg/+, skg/skg, and skg/- mice
Sun Aug 15 00:00:00 EDT 2010
Self-organized criticality theory of autoimmunity.
PloS one
Tsumiyama K,Miyazaki Y,Shiozawa S
MM7501 was used in flow cytometry to discuss causes of autoimmunity
Thu Dec 31 00:00:00 EST 2009
Distinct regulation of integrin-dependent T cell conjugate formation and NF-kappa B activation by the adapter protein ADAP.
Journal of immunology (Baltimore, Md. : 1950)
Burbach BJ,Srivastava R,Medeiros RB,O'Gorman WE,Peterson EJ,Shimizu Y
MM7501 was used in flow cytometry to elucidate how adhesion and degranulation-promoting adapter protein controls integrin adhesive function and NF-kappaB transcription factor activation.
Wed Oct 01 00:00:00 EDT 2008
Lactobacillus acidophilus strain L-92 induces apoptosis of antigen-stimulated T cells by modulating dendritic cell function.
Immunobiology
Kanzato H,Fujiwara S,Ise W,Kaminogawa S,Sato R,Hachimura S
MM7501 was used in flow cytometry to study the effect of Lactobacillus acidophilus strain L-92 on antigen-stimulated T cell responses in vitro and in vivo
Tue Feb 03 00:00:00 EST 2009
Periportal and sinusoidal liver dendritic cells suppressing T helper type 1-mediated hepatitis.
Gut
Watanabe T,Katsukura H,Chiba T,Kita T,Wakatsuki Y
MM7501 was used in flow cytometry to test if periportal and sinusoidal liver dendritic cells suppress T helper type 1-mediated hepatitis
Mon Oct 01 00:00:00 EDT 2007
Induction and role of regulatory CD4+CD25+ T cells in tolerance to the transgene product following hepatic in vivo gene transfer.
Blood
Cao O,Dobrzynski E,Wang L,Nayak S,Mingle B,Terhorst C,Herzog RW
MM7501 was used in flow cytometry to provide the first definitive evidence for the induction of transgene product-specific CD4+CD25+ T regulatory cells by in vivo gene transfer
Wed Aug 15 00:00:00 EDT 2007
Transmaternal exposure to bisphenol a modulates the development of oral tolerance.
Pediatric research
Ohshima Y,Yamada A,Tokuriki S,Yasutomi M,Omata N,Mayumi M
MM7501 was used in flow cytometry to test if transmaternal exposure to bisphenol A influences the development of oral tolerance
Sun Jul 01 00:00:00 EDT 2007
IgE enhances specific antibody and T-cell responses in mice overexpressing CD23.
Scandinavian journal of immunology
Carlsson F,Hjelm F,Conrad DH,Heyman B
MM7501 was used in flow cytometry to investigate if IgE enhances antibody and T-cell responses in IgE receptor CD23 transgenic animals or if CD23's downregulatory effect precludes IgE-mediated enhancement
Fri Sep 14 00:00:00 EDT 2007
Antigen and lipopolysaccharide play synergistic roles in the effector phase of airway inflammation in mice.
The American journal of pathology
Jung YW,Schoeb TR,Weaver CT,Chaplin DD
MM7501 was used in flow cytometry to clarify the role of innate immune signals in the development of airway inflammation
Mon May 01 00:00:00 EDT 2006
G protein-coupled receptor 83 overexpression in naive CD4+CD25- T cells leads to the induction of Foxp3+ regulatory T cells in vivo.
Journal of immunology (Baltimore, Md. : 1950)
Hansen W,Loser K,Westendorf AM,Bruder D,Pfoertner S,Siewert C,Huehn J,Beissert S,Buer J
MM7501 was used in flow cytometry to investigate the role of GPR83 in the peripheral generation of Foxp3+ T regulatory cells
Sat Jul 01 00:00:00 EDT 2006
The role of TCR engagement and activation-induced cell death in sepsis-induced T cell apoptosis.
Journal of immunology (Baltimore, Md. : 1950)
Unsinger J,Herndon JM,Davis CG,Muenzer JT,Hotchkiss RS,Ferguson TA
MM7501 was used to test if proliferation and AICD are necessary for apoptotic cell death in sepsis using normal and TCR transgenic mice
Fri Dec 01 00:00:00 EST 2006
Systemic protein delivery by muscle-gene transfer is limited by a local immune response.
Blood
Wang L,Dobrzynski E,Schlachterman A,Cao O,Herzog RW
MM7501 was used in immunohistochemistry - frozen section to show immune-mediated elimination of expression on muscle-directed adeno-associated viral-2 gene transfer
Wed Jun 01 00:00:00 EDT 2005
Suppression of CD4+ T cell activation by a novel inhibitor of Src family kinases.
International immunopharmacology
McRae BL,Wallace C,Dixon KF,Roux A,Mohan S,Jia Y,Presky DH,Tracey DE,Hirst GC
MM7501 was used in flow cytometry to study the suppression of CD4+ T cell activation by a novel small molecule inhibitor of Src family kinases
Fri Apr 01 00:00:00 EST 2005
Retinoid x receptor agonists increase bcl2a1 expression and decrease apoptosis of naive T lymphocytes.
Journal of immunology (Baltimore, Md. : 1950)
Rasooly R,Schuster GU,Gregg JP,Xiao JH,Chandraratna RA,Stephensen CB
MM7501 was used in flow cytometry to identify RXR-responsive genes in T cells
Thu Dec 15 00:00:00 EST 2005
An optimized CD4 T-cell response can control productive and latent gammaherpesvirus infection.
Journal of virology
Sparks-Thissen RL,Braaten DC,Kreher S,Speck SH,Virgin HW
MM7501 was used in flow cytometry to test if CD4 T cells control gamma HV68 infection in the absence of CD8 T cells or B cells
Thu Jul 01 00:00:00 EDT 2004
Intranasal treatment with ovalbumin but not the major T cell epitope ovalbumin 323-339 generates interleukin-10 secreting T cells and results in the induction of allergen systemic tolerance.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology
Barbey C,Donatelli-Dufour N,Batard P,Corradin G,Spertini F
MM7501 was used in flow cytometry to discuss the consequences of nasal administration of major peptide T cell epitopes
Thu Apr 01 00:00:00 EST 2004
Antigenic activation of Th1 cells in the gastric mucosa enhances dysregulated apoptosis and turnover of the epithelial cells.
Biochemical and biophysical research communications
Yamori M,Yoshida M,Watanabe T,Shirai Y,Iizuka T,Kita T,Wakatsuki Y
MM7501 was used in flow cytometry to investigate the role of T cells in the remodeling of gastric epithelium of Helicobacter pylori-infected mice
Fri Apr 16 00:00:00 EDT 2004
Visualization of the genesis and fate of isotype-switched B cells during a primary immune response.
The Journal of experimental medicine
Pape KA,Kouskoff V,Nemazee D,Tang HL,Cyster JG,Tze LE,Hippen KL,Behrens TW,Jenkins MK
MM7501 was used in flow cytometry to elucidate the history and localization of isotype-switched B cells.
Mon Jun 16 00:00:00 EDT 2003
Preferential accumulation of antigen-specific effector CD4 T cells at an antigen injection site involves CD62E-dependent migration but not local proliferation.
The Journal of experimental medicine
Reinhardt RL,Bullard DC,Weaver CT,Jenkins MK
MM7501 was used in flow cytometry to study the migration of antigen-specific T cells to nonlymphoid tissues.
Mon Mar 17 00:00:00 EST 2003
Direct expansion of functional CD25+ CD4+ regulatory T cells by antigen-processing dendritic cells.
The Journal of experimental medicine
Yamazaki S,Iyoda T,Tarbell K,Olson K,Velinzon K,Inaba K,Steinman RM
MM7501 was used in flow cytometry to study antigen-specific expansion of CD25+ CD4+ T cells.
Mon Jul 21 00:00:00 EDT 2003
Burn injury promotes antigen-driven Th2-type responses in vivo.
Journal of immunology (Baltimore, Md. : 1950)
Guo Z,Kavanagh E,Zang Y,Dolan SM,Kriynovich SJ,Mannick JA,Lederer JA
MM7501 was used in flow cytometry to explore how injury skews T cell responses.
Wed Oct 15 00:00:00 EDT 2003
Type I interferons produced by dendritic cells promote their phenotypic and functional activation.
Blood
Montoya M,Schiavoni G,Mattei F,Gresser I,Belardelli F,Borrow P,Tough DF
MM7501 was used in flow cytometry to test the effects of IFN on dendritic cells.
Wed May 01 00:00:00 EDT 2002
In situ analysis reveals physical interactions between CD11b+ dendritic cells and antigen-specific CD4 T cells after subcutaneous injection of antigen.
Journal of immunology (Baltimore, Md. : 1950)
Ingulli E,Ulman DR,Lucido MM,Jenkins MK
MM7501 was used in flow cytometry to visualize the physical interactions between dendritic cell subsets and naive antigen-specific CD4 T cells in the lymph node.
Sun Sep 01 00:00:00 EDT 2002
Signal transducer and activator of transcription 6 controls chemokine production and T helper cell type 2 cell trafficking in allergic pulmonary inflammation.
The Journal of experimental medicine
Mathew A,MacLean JA,DeHaan E,Tager AM,Green FH,Luster AD
MM7501 was used in flow cytometry to investigate how Th2 cells are recruited to the lungs during allergic asthma.
Mon May 07 00:00:00 EDT 2001
Single-cell analysis of signal transduction in CD4 T cells stimulated by antigen in vivo.
Proceedings of the National Academy of Sciences of the United States of America
Zell T,Khoruts A,Ingulli E,Bonnevier JL,Mueller DL,Jenkins MK
MM7501 was used in flow cytometry to investigate signaling events in individual CD4 T cells after antigen recognition.
Tue Sep 11 00:00:00 EDT 2001
Single-cell analysis of signal transduction in CD4 T cells stimulated by antigen in vivo.
Proceedings of the National Academy of Sciences of the United States of America
Zell T,Khoruts A,Ingulli E,Bonnevier JL,Mueller DL,Jenkins MK
MM7501 was used in flow cytometry to investigate signaling events in individual CD4 T cells after antigen recognition.
Tue Sep 11 00:00:00 EDT 2001
Survival and cytokine polarization of naive CD4(+) T cells in vitro is largely dependent on exogenous cytokines.
European journal of immunology
Ben-Sasson SZ,Makedonski K,Hu-Li J,Paul WE
MM7501 was used in flow cytometry to study the effect of IL-4 on T cell priming.
Mon May 01 00:00:00 EDT 2000