Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In Western blot applications, this antibody detects a band at ~87kDa. A suggested positive control is Jurkat cell lysate.
PA5-19916 can be used with blocking peptide PEP-0042.
Phosphorylation of serine residues on the I-kappa-B proteins by IKK kinases, including IKBKB, marks them for destruction via the ubiquitination pathway, allowing activation of the NF-kappa-B complex. Inhibitor of nuclear factor kappa-B kinase subunit beta (IKK beta) is a serine kinase that plays an essential role in the NF-kaapa-B signaling pathway which is activated by mulitple stimuli such as inflammatory cytokines, bacterial, or viral products, DNA damages, or other cellular stresses. IKK beta acts as part of the canonical IKK complex in the conventional pathway of NF-kappa-B activation and phosphorylates inhibitors of NF-kappa-B on 2 critical serine residues. These modifications allow polyubiquitination of the inhibitors and subsequent degradation by the proteasome. In turn, free NF-kappa-B is translocated into the nucleus and activates the transcription of hundreds of genes involved in immune response, growth control, or protection against apoptosis. Mutations affecting the IKK beta gene can result in Immunodeficiency 15 (IMD15).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ40509; I-kappa-B kinase 2; I-kappa-B-kinase beta; IKK-B; IKK2; Inhibitor of nuclear factor kappa-B kinase subunit beta; MGC131801; NFKBIKB; Nuclear factor NF-kappa-B inhibitor kinase beta; Serine/threonine protein kinase IKBKB
Gene Aliases: IKBKB; IKK-beta; IKK2; IKKB; IMD15; NFKBIKB
UniProt ID: (Human) O14920
Entrez Gene ID: (Human) 3551
Molecular Function: non-receptor serine/threonine protein kinase protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support