Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PBRM1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_018313.4 | NP_060783.3 | - | - | - |
XM_005265279.4 | XP_005265336.1 | - | - | - | |
XM_005265280.3 | XP_005265337.1 | - | - | - | |
XM_005265282.3 | XP_005265339.1 | - | - | - | |
XM_005265283.3 | XP_005265340.1 | - | - | - | |
XM_011533900.2 | XP_011532202.1 | - | - | - | |
XM_011533902.2 | XP_011532204.1 | - | - | - | |
XM_011533903.2 | XP_011532205.1 | - | - | - | |
XM_017006725.1 | XP_016862214.1 | - | - | - | |
XM_017006726.1 | XP_016862215.1 | - | - | - | |
XM_017006727.1 | XP_016862216.1 | - | - | - | |
XM_017006728.1 | XP_016862217.1 | - | - | - | |
XM_017006729.1 | XP_016862218.1 | - | - | - | |
XM_017006730.1 | XP_016862219.1 | - | - | - | |
XM_017006731.1 | XP_016862220.1 | - | - | - | |
XM_017006732.1 | XP_016862221.1 | 6954461 | c.4940G>A | p.R1647H | |
6332433 | c.5000G>T | p.W1667L | |||
6506752 | c.4973delC | p.H1659fs*14 | |||
5611348 | c.5024_5025GG>AA | p.R1675Q | |||
6940305 | c.5008C>T | p.R1670* | |||
6919686 | c.5016G>C | p.L1672F | |||
5646385 | c.5016_5017insT | p.M1673fs*>18 | |||
480304 | c.5009G>C | p.R1670P | |||
3596166 | c.5014T>A | p.L1672M | |||
6937647 | c.5041C>T | p.R1681C | |||
249530 | c.4925_4926delAA | p.K1642fs*>48 | |||
5095294 | c.5023C>T | p.R1675W | |||
3775235 | c.4959G>A | p.L1653L | |||
5463284 | c.5009G>A | p.R1670Q | |||
XM_017006733.1 | XP_016862222.1 | - | - | - | |
XM_017006734.1 | XP_016862223.1 | - | - | - | |
XM_017006735.1 | XP_016862224.1 | - | - | - | |
XM_017006736.1 | XP_016862225.1 | - | - | - | |
XM_017006737.1 | XP_016862226.1 | - | - | - | |
XM_017006738.1 | XP_016862227.1 | - | - | - | |
XM_017006739.1 | XP_016862228.1 | - | - | - | |
XM_017006740.1 | XP_016862229.1 | - | - | - | |
XM_017006741.1 | XP_016862230.1 | - | - | - | |
XM_017006742.1 | XP_016862231.1 | - | - | - | |
XM_017006743.1 | XP_016862232.1 | - | - | - | |
XM_017006744.1 | XP_016862233.1 | - | - | - | |
XM_017006745.1 | XP_016862234.1 | - | - | - | |
XM_017006746.1 | XP_016862235.1 | - | - | - | |
XM_017006747.1 | XP_016862236.1 | - | - | - | |
XM_017006748.1 | XP_016862237.1 | - | - | - | |
XM_017006749.1 | XP_016862238.1 | - | - | - | |
XM_017006750.1 | XP_016862239.1 | - | - | - | |
XM_017006751.1 | XP_016862240.1 | - | - | - | |
XM_017006752.1 | XP_016862241.1 | - | - | - | |
XM_017006753.1 | XP_016862242.1 | - | - | - | |
XM_017006754.1 | XP_016862243.1 | - | - | - | |
XM_017006755.1 | XP_016862244.1 | - | - | - | |
XM_017006756.1 | XP_016862245.1 | - | - | - | |
XM_017006757.1 | XP_016862246.1 | - | - | - | |
XM_017006758.1 | XP_016862247.1 | - | - | - | |
XM_017006759.1 | XP_016862248.1 | - | - | - | |
XM_017006760.1 | XP_016862249.1 | - | - | - | |
XM_017006761.1 | XP_016862250.1 | - | - | - | |
XM_017006762.1 | XP_016862251.1 | - | - | - | |
XM_017006763.1 | XP_016862252.1 | - | - | - | |
XM_017006764.1 | XP_016862253.1 | 6919685 | c.4695G>C | p.L1565F | |
6506751 | c.4652delC | p.H1552fs*14 | |||
1169603 | c.4642_4642delA | p.S1548fs*18 | |||
1169566 | c.4661_4680del20 | p.M1555fs*23 | |||
6332432 | c.4679G>T | p.W1560L | |||
6940304 | c.4687C>T | p.R1563* | |||
5095293 | c.4702C>T | p.R1568W | |||
6954460 | c.4619G>A | p.R1540H | |||
3596165 | c.4693T>A | p.L1565M | |||
5463283 | c.4688G>A | p.R1563Q | |||
1169589 | c.4693_4705del13 | p.L1565fs*11 | |||
6937646 | c.4720C>T | p.R1574C | |||
5646384 | c.4695_4696insT | p.M1566fs*>18 | |||
3775234 | c.4638G>A | p.L1546L | |||
480303 | c.4688G>C | p.R1563P | |||
5611347 | c.4703_4704GG>AA | p.R1568Q | |||
XM_017006765.1 | XP_016862254.1 | - | - | - | |
XM_017006766.1 | XP_016862255.1 | - | - | - | |
GenBank | AF177387.1 | AAG48933.1 | - | - | - |
AF197569.1 | AAG34760.1 | - | - | - | |
AF225870.1 | AAG48939.1 | - | - | - | |
AF225871.1 | AAG48940.1 | - | - | - | |
AF225872.1 | AAG48941.1 | - | - | - | |
AK001507.1 | BAA91728.1 | - | - | - | |
AY281068.1 | AAP34197.1 | - | - | - | |
BC015323.1 | AAH15323.1 | - | - | - | |
BC089409.1 | AAH89409.1 | - | - | - | |
BC115009.1 | AAI15010.1 | - | - | - | |
BC115010.1 | - | - | - | ||
BC115011.1 | AAI15012.1 | - | - | - | |
BC116450.1 | - | - | - | ||
BC129934.1 | - | - | - | ||
BC129935.1 | - | - | - | ||
DA039085.1 | - | - | - | ||
ENST00000337303.0 | - | - | - |
Literature Links: |
PBRM1 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |