Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MYB | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001130172.1 | NP_001123644.1 | - | - | - |
NM_001130173.1 | NP_001123645.1 | 4186544 | c.340C>T | p.R114C | |
1487252 | c.308T>C | p.V103A | |||
1620950 | c.325A>C | p.K109Q | |||
5731878 | c.335C>T | p.P112L | |||
4704273 | c.341G>A | p.R114H | |||
NM_001161656.1 | NP_001155128.1 | - | - | - | |
NM_001161657.1 | NP_001155129.1 | - | - | - | |
NM_001161658.1 | NP_001155130.1 | - | - | - | |
NM_001161659.1 | NP_001155131.1 | - | - | - | |
NM_001161660.1 | NP_001155132.1 | - | - | - | |
NM_005375.3 | NP_005366.2 | 4186543 | c.340C>T | p.R114C | |
4704272 | c.341G>A | p.R114H | |||
1620949 | c.325A>C | p.K109Q | |||
5731877 | c.335C>T | p.P112L | |||
1487251 | c.308T>C | p.V103A | |||
NR_134958.1 | - | - | - | ||
NR_134959.1 | - | - | - | ||
NR_134960.1 | - | - | - | ||
NR_134961.1 | - | - | - | ||
NR_134962.1 | - | - | - | ||
NR_134963.1 | - | - | - | ||
NR_134964.1 | - | - | - | ||
NR_134965.1 | - | - | - | ||
GenBank | AF104863.1 | AAC96326.1 | - | - | - |
AJ606317.1 | CAE55168.1 | - | - | - | |
AJ606318.1 | CAE55169.1 | - | - | - | |
AJ606319.1 | CAE55170.1 | - | - | - | |
AJ606320.1 | CAE55171.1 | - | - | - | |
AJ606321.1 | CAE55172.1 | - | - | - | |
AJ606322.1 | CAE55173.1 | - | - | - | |
AJ606323.1 | CAE55174.1 | - | - | - | |
AJ606324.1 | CAE55175.1 | - | - | - | |
AJ616235.1 | CAE82649.1 | - | - | - | |
AJ616795.1 | - | - | - | ||
AY787443.1 | - | - | - | ||
AY787444.1 | - | - | - | ||
AY787445.1 | - | - | - | ||
AY787446.1 | - | - | - | ||
AY787447.1 | - | - | - | ||
AY787448.1 | - | - | - | ||
AY787449.1 | - | - | - | ||
AY787450.1 | - | - | - | ||
AY787451.1 | - | - | - | ||
AY787452.1 | - | - | - | ||
AY787453.1 | - | - | - | ||
AY787454.1 | - | - | - | ||
AY787455.1 | - | - | - | ||
AY787456.1 | - | - | - | ||
AY787457.1 | - | - | - | ||
AY787458.1 | - | - | - | ||
AY787459.1 | - | - | - | ||
AY787460.1 | - | - | - | ||
AY787461.1 | - | - | - | ||
AY787462.1 | - | - | - | ||
AY787463.1 | - | - | - | ||
AY787464.1 | - | - | - | ||
AY787465.1 | - | - | - | ||
AY787466.1 | - | - | - | ||
AY787467.1 | - | - | - | ||
AY787468.1 | - | - | - | ||
AY787469.1 | - | - | - | ||
AY787470.1 | - | - | - | ||
AY787471.1 | - | - | - | ||
AY787472.1 | - | - | - | ||
AY787473.1 | - | - | - | ||
AY787474.1 | - | - | - | ||
AY787475.1 | - | - | - | ||
BC064955.1 | AAH64955.1 | - | - | - | |
ENST00000367814.0 | - | - | - | ||
M13665.1 | AAA52030.1 | - | - | - | |
M13666.1 | AAA52031.1 | - | - | - | |
M15024.1 | AAA52032.1 | - | - | - | |
X52125.1 | CAA36371.1 | - | - | - | |
X52126.1 | CAA36372.1 | - | - | - |
Literature Links: |
MYB PubMed Links |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
Set Membership: |
Less than 200 bp |