Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GDNF | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_000514.3 | NP_000505.1 | 1754265 | c.500G>C | p.R167T |
222057 | c.354T>A | p.C118* | |||
2688718 | c.420G>A | p.L140L | |||
1437399 | c.468C>T | p.Y156Y | |||
161238 | c.364G>C | p.A122P | |||
4347864 | c.406A>G | p.T136A | |||
1671600 | c.416A>G | p.E139G | |||
172198 | c.447C>T | p.C149C | |||
6020207 | c.491C>G | p.S164C | |||
1208072 | c.466T>C | p.Y156H | |||
3721917 | c.426T>A | p.F142L | |||
1235070 | c.350G>A | p.G117D | |||
6170941 | c.478T>C | p.L160L | |||
3410260 | c.439G>A | p.G147S | |||
737856 | c.491C>T | p.S164F | |||
5554651 | c.440G>A | p.G147D | |||
3616030 | c.455C>T | p.A152V | |||
3616034 | c.396G>A | p.L132L | |||
2156663 | c.379G>A | p.V127I | |||
4999474 | c.457G>T | p.E153* | |||
2688713 | c.464C>T | p.T155M | |||
7238617 | c.393T>C | p.G131G | |||
1437401 | c.357C>A | p.V119V | |||
3941308 | c.457G>A | p.E153K | |||
1716946 | c.347_359del13 | p.R116fs*6 | |||
3616036 | c.358T>G | p.L120V | |||
6103710 | c.459G>T | p.E153D | |||
6170939 | c.398G>T | p.G133V | |||
3749928 | c.425T>G | p.F142C | |||
1253166 | c.465G>A | p.T155T | |||
1437397 | c.487T>G | p.L163V | |||
329036 | c.431A>C | p.Y144S | |||
5120626 | c.351T>A | p.G117G | |||
7425530 | c.431A>T | p.Y144F | |||
3616032 | c.412G>A | p.E138K | |||
NM_001190468.1 | NP_001177397.1 | 1716945 | c.398_410del13 | p.R133fs*6 | |
4810477 | c.415G>C | p.A139P | |||
1754264 | c.551G>C | p.R184T | |||
4999473 | c.508G>T | p.E170* | |||
2688714 | c.498C>T | p.C166C | |||
1437398 | c.519C>T | p.Y173Y | |||
3941307 | c.508G>A | p.E170K | |||
4347863 | c.457A>G | p.T153A | |||
3616035 | c.409T>G | p.L137V | |||
6020206 | c.542C>G | p.S181C | |||
1208071 | c.517T>C | p.Y173H | |||
3749927 | c.476T>G | p.F159C | |||
6170940 | c.529T>C | p.L177L | |||
1235069 | c.401G>A | p.G134D | |||
7238616 | c.444T>C | p.G148G | |||
2688717 | c.471G>A | p.L157L | |||
3410259 | c.490G>A | p.G164S | |||
5554650 | c.491G>A | p.G164D | |||
1150063 | c.542C>T | p.S181F | |||
3616031 | c.463G>A | p.E155K | |||
1671599 | c.467A>G | p.E156G | |||
2156662 | c.430G>A | p.V144I | |||
2688712 | c.515C>T | p.T172M | |||
1437396 | c.538T>G | p.L180V | |||
1437400 | c.408C>A | p.V136V | |||
6103709 | c.510G>T | p.E170D | |||
3721916 | c.477T>A | p.F159L | |||
6170938 | c.449G>T | p.G150V | |||
1253165 | c.516G>A | p.T172T | |||
7425529 | c.482A>T | p.Y161F | |||
3616029 | c.506C>T | p.A169V | |||
3616033 | c.447G>A | p.L149L | |||
NM_001190469.1 | NP_001177398.1 | - | - | - | |
NM_001278098.1 | NP_001265027.1 | - | - | - | |
NM_199231.2 | NP_954701.1 | - | - | - | |
XM_011514030.2 | XP_011512332.1 | - | - | - | |
XM_017009337.1 | XP_016864826.1 | - | - | - | |
GenBank | AF053748.1 | AAD43139.1 | - | - | - |
AY052832.1 | AAL11017.1 | - | - | - | |
BC069119.1 | AAH69119.1 | - | - | - | |
BC069369.1 | AAH69369.1 | - | - | - | |
BC128108.1 | - | - | - | ||
BC128109.1 | - | - | - | ||
CR541923.1 | CAG46721.1 | - | - | - | |
DQ235474.1 | - | - | - | ||
ENST00000326524.0 | - | - | - | ||
KU178093.1 | - | - | - | ||
KU178094.1 | - | - | - |
Literature Links: |
GDNF PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
Set Membership: |
Less than 200 bp |