Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
SMUG1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001243787.1 | NP_001230716.1 | 1606334 | c.380T>G | p.L127R |
1726371 | c.383G>C | p.G128A | |||
6785795 | c.310G>A | p.V104I | |||
1677026 | c.419G>A | p.R140Q | |||
5567365 | c.400T>C | p.S134P | |||
4957553 | c.364C>T | p.P122S | |||
4915045 | c.300A>C | p.E100D | |||
3368866 | c.398A>T | p.Q133L | |||
6879192 | c.395C>T | p.P132L | |||
6785802 | c.418C>T | p.R140* | |||
4807873 | c.370C>T | p.R124* | |||
NM_001243788.1 | NP_001230717.1 | - | - | - | |
NM_001243789.1 | NP_001230718.1 | 5567366 | c.400T>C | p.S134P | |
3704250 | c.380T>G | p.L127R | |||
3368867 | c.398A>T | p.Q133L | |||
4957554 | c.364C>T | p.P122S | |||
4915046 | c.300A>C | p.E100D | |||
4807874 | c.370C>T | p.R124* | |||
6879193 | c.395C>T | p.P132L | |||
7309060 | c.383G>C | p.G128A | |||
6785796 | c.310G>A | p.V104I | |||
NM_001243790.1 | NP_001230719.1 | - | - | - | |
NM_001243791.1 | NP_001230720.1 | - | - | - | |
NM_014311.2 | NP_055126.1 | - | - | - | |
NR_045039.1 | - | - | - | ||
XM_006719319.3 | XP_006719382.1 | - | - | - | |
XM_006719320.3 | XP_006719383.1 | - | - | - | |
XM_006719321.3 | XP_006719384.1 | - | - | - | |
XM_006719322.3 | XP_006719385.1 | - | - | - | |
XM_011538109.2 | XP_011536411.1 | - | - | - | |
XM_011538110.2 | XP_011536412.1 | - | - | - | |
XM_011538111.2 | XP_011536413.1 | - | - | - | |
XM_011538112.2 | XP_011536414.1 | - | - | - | |
XM_011538113.2 | XP_011536415.1 | - | - | - | |
XM_011538114.2 | XP_011536416.1 | - | - | - | |
XM_011538115.2 | XP_011536417.1 | - | - | - | |
XM_011538116.2 | XP_011536418.1 | - | - | - | |
XM_011538117.2 | XP_011536419.1 | - | - | - | |
XM_011538118.2 | XP_011536420.1 | - | - | - | |
XM_011538119.2 | XP_011536421.1 | - | - | - | |
XM_011538120.2 | XP_011536422.1 | - | - | - | |
XM_011538121.2 | XP_011536423.1 | - | - | - | |
XM_011538122.2 | XP_011536424.1 | - | - | - | |
XM_011538123.2 | XP_011536425.1 | - | - | - | |
XM_017019113.1 | XP_016874602.1 | - | - | - | |
XM_017019114.1 | XP_016874603.1 | - | - | - | |
XM_017019115.1 | XP_016874604.1 | - | - | - | |
XM_017019116.1 | XP_016874605.1 | - | - | - | |
XM_017019117.1 | XP_016874606.1 | - | - | - | |
XM_017019118.1 | XP_016874607.1 | - | - | - | |
XM_017019119.1 | XP_016874608.1 | - | - | - | |
XM_017019120.1 | XP_016874609.1 | - | - | - | |
GenBank | AF125182.1 | AAD17301.1 | - | - | - |
AK001235.1 | BAA91571.1 | - | - | - | |
AK001789.1 | - | - | - | ||
AK091468.1 | BAC03670.1 | - | - | - | |
AK222473.1 | BAD96193.1 | - | - | - | |
AK290274.1 | - | - | - | ||
BC000417.2 | AAH00417.1 | - | - | - | |
BC088352.1 | AAH88352.1 | - | - | - | |
BC105607.1 | - | - | - | ||
BG706635.1 | - | - | - | ||
BG764502.1 | - | - | - | ||
BU195802.1 | - | - | - | ||
CD245413.1 | - | - | - | ||
DA430734.1 | - | - | - | ||
ENST00000337581.0 | - | - | - | ||
KU178578.1 | - | - | - | ||
KU178579.1 | - | - | - |
Literature Links: |
SMUG1 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |