Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PIK3CA | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_006218.3 | NP_006209.2 | 3408452 | c.308A>G | p.E103G |
5029069 | c.316_321delGGCAAC | p.G106_N107delGN | |||
24711 | c.335T>A | p.I112N | |||
5752504 | c.333_338delGATCCT | p.K111_L113>N | |||
27497 | c.323G>A | p.R108H | |||
6986613 | c.325G>A | p.E109K | |||
7347215 | c.289_290insTG | p.P97fs*4 | |||
6984365 | c.344G>A | p.R115Q | |||
5347117 | c.225A>C | p.Q75H | |||
27493 | c.277C>T | p.R93W | |||
7403490 | c.310C>G | p.P104A | |||
14052 | c.332A>G | p.K111R | |||
4971083 | c.328_330delGAA | p.E110delE | |||
1041458 | c.304_306delATT | p.I102delI | |||
6941709 | c.305_307delTTG | p.I102_E103>K | |||
3695922 | c.241G>T | p.E81* | |||
6438099 | c.305T>C | p.I102T | |||
6191568 | c.274C>T | p.L92F | |||
5954758 | c.252T>C | p.D84D | |||
7346036 | c.313delG | p.V105fs*1 | |||
163485 | c.311C>T | p.P104L | |||
6438094 | c.295_296delTT | p.L99fs*4 | |||
6163880 | c.260G>C | p.R87T | |||
6961565 | c.314_320TAGGCAA>G | p.V105_N107>G | |||
6911319 | c.325_326insAAGAAA | p.K111_I112insEK | |||
86041 | c.278G>A | p.R93Q | |||
6976103 | c.304A>T | p.I102F | |||
6475618 | c.325G>T | p.E109* | |||
27505 | c.333G>T | p.K111N | |||
747 | c.311C>G | p.P104R | |||
29641 | c.316_324del | p.G106_R108del | |||
6923933 | c.346G>A | p.E116K | |||
5752260 | c.331_336delAAGATC | p.K111_I112delKI | |||
1484855 | c.307_312delGAACCA | p.E103_P104delEP | |||
12580 | c.333G>C | p.K111N | |||
746 | c.263G>A | p.R88Q | |||
6950313 | c.322_334>G | p.R108_I112>V | |||
24710 | c.325_327delGAA | p.E109del | |||
6475598 | c.340A>G | p.N114D | |||
5020505 | c.341A>G | p.N114S | |||
3408450 | c.269G>A | p.C90Y | |||
6577274 | c.313_324del12 | p.V105_R108delVGNR | |||
7347213 | c.277_278insCCATAGAGA | p.L92_R93insPIE | |||
21448 | c.337_339delCTC | p.L113del | |||
1239841 | c.309A>G | p.E103E | |||
7412594 | c.277C>G | p.R93G | |||
1041466 | c.335_337delTCC | p.L113delL | |||
166154 | c.337_342delCTCAAT | p.L113_N114delLN | |||
7346044 | c.329_331delAAA | p.K111delK | |||
7346034 | c.310_311delCC | p.P104fs*5 | |||
304777 | c.248T>A | p.F83Y | |||
163484 | c.238G>A | p.E80K | |||
445998 | c.331_333delAAG | p.K111delK | |||
7342608 | c.297A>T | p.L99F | |||
445992 | c.316_324delGGCAACCGT | p.G106_R108delGNR | |||
6908894 | c.334_339delATCCTC | p.I112_L113delIL | |||
5547415 | c.341_343delATC | p.N114delN | |||
750 | c.332_334delAGA | p.K111del | |||
7347342 | c.292_298delTTTTTAA | p.F98fs*2 | |||
6923573 | c.320A>T | p.N107I | |||
1666660 | c.310C>A | p.P104T | |||
7339127 | c.302_304delTAA | p.V101del | |||
5515965 | c.250G>A | p.D84N | |||
7347211 | c.273_274insACCGA | p.L92fs*10 | |||
214489 | c.309_317delACCAGTAGG | p.E103_G106>D | |||
6475668 | c.327A>G | p.E109E | |||
5736834 | c.244delT | p.F83fs*17 | |||
6475604 | c.263G>T | p.R88L | |||
27499 | c.321_323delCCG | p.R108del | |||
3408448 | c.268T>G | p.C90G | |||
6573714 | c.213A>G | p.V71V | |||
1041462 | c.320A>G | p.N107S | |||
7346712 | c.286C>G | p.Q96E | |||
166340 | c.318C>A | p.G106G | |||
3737049 | c.253G>A | p.E85K | |||
6865935 | c.330_332delAAA | p.K111del | |||
1235453 | c.334A>T | p.I112F | |||
6940781 | c.309_314delACCAGT | p.P104_V105delPV | |||
7346042 | c.308_310delAAC | p.E103_P104>A | |||
6191573 | c.280C>T | p.L94F | |||
4971439 | c.304_315del12 | p.I102_V105delIEPV | |||
6191359 | c.278G>C | p.R93P | |||
5945917 | c.249T>G | p.F83L | |||
7346032 | c.323_334del12 | p.R108_I112>L | |||
13570 | c.331A>G | p.K111E | |||
1645488 | c.316G>A | p.G106S | |||
6438096 | c.321C>A | p.N107K | |||
5346971 | c.248T>C | p.F83S | |||
445993 | c.316_324del | p.G106_R108del | |||
6913225 | c.247_249TTT>AAG | p.F83K | |||
51259 | c.341_342insCCTCAA | p.N114_R115insLN | |||
6145 | c.328G>A | p.E110K | |||
749 | c.323G>C | p.R108P | |||
125368 | c.344G>T | p.R115L | |||
6746431 | c.316G>T | p.G106C | |||
7450115 | c.275_276ins17 | p.R93fs*13 | |||
6475743 | c.307G>A | p.E103K | |||
7347340 | c.281_284delTTTT | p.F95fs*4 | |||
27502 | c.241G>A | p.E81K | |||
6906148 | c.317_328del12 | p.G106_E109del | |||
6218467 | c.320A>C | p.N107T | |||
7347209 | c.272_273insT | p.L92fs*12 | |||
27498 | c.311_316delCAGTAG | p.P104_G106>R | |||
1235451 | c.317G>A | p.G106D | |||
6545055 | c.319A>T | p.N107Y | |||
3364898 | c.214A>G | p.S72G | |||
6941830 | c.232G>A | p.E78K | |||
1041460 | c.315_320delAGGCAA | p.G106_N107delGN | |||
1753026 | c.269G>C | p.C90S | |||
4517286 | c.310_311CC>TT | p.P104L | |||
166339 | c.317G>C | p.G106A | |||
729829 | c.323G>T | p.R108L | |||
5347052 | c.242A>C | p.E81A | |||
6983144 | c.290C>A | p.P97Q | |||
27490 | c.333_335delGAT | p.K111_I112>N | |||
6851672 | c.305T>A | p.I102N | |||
6963571 | c.344G>C | p.R115P | |||
7346038 | c.323_325delGTG | p.R108_E109>Q | |||
6978772 | c.322C>A | p.R108S | |||
6438095 | c.322C>T | p.R108C | |||
6960410 | c.331A>C | p.K111Q | |||
4832940 | c.268T>C | p.C90R | |||
6475767 | c.225A>G | p.Q75Q | |||
7346030 | c.310_318delCCAGTAGGC | p.P104_G106delPVG | |||
6914463 | c.278G>T | p.R93L | |||
86046 | c.316G>C | p.G106R | |||
6746448 | c.224A>C | p.Q75P | |||
6976719 | c.317_320GCAA>TCAG | p.G106_N107>VS | |||
748 | c.317G>T | p.G106V | |||
39166 | c.223C>G | p.Q75E | |||
XM_006713658.3 | XP_006713721.1 | - | - | - | |
XM_011512894.2 | XP_011511196.1 | - | - | - | |
GenBank | AB210020.1 | - | - | - | |
AK292940.1 | - | - | - | ||
AK302608.1 | - | - | - | ||
BC113601.1 | - | - | - | ||
BC113603.1 | - | - | - | ||
BX473473.1 | - | - | - | ||
U79143.1 | AAB39753.1 | - | - | - | |
Z29090.1 | CAA82333.1 | - | - | - |
Literature Links: |
PIK3CA PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |