Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACNA1E | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_000721.3 | NP_000712.2 | 1336829 | c.1723C>T | p.R575W |
3376861 | c.1688G>T | p.G563V | |||
6541385 | c.1682G>T | p.R561I | |||
424744 | c.1714C>T | p.R572* | |||
279524 | c.1710C>A | p.V570V | |||
6894690 | c.1734A>C | p.R578S | |||
463519 | c.1665G>T | p.V555V | |||
4925410 | c.1663G>A | p.V555M | |||
4452282 | c.1747A>C | p.T583P | |||
1199274 | c.1691C>T | p.T564M | |||
4668484 | c.1692G>A | p.T564T | |||
900534 | c.1688G>A | p.G563D | |||
6541390 | c.1715G>T | p.R572L | |||
5527931 | c.1682G>A | p.R561K | |||
4569411 | c.1745T>A | p.I582K | |||
219242 | c.1673C>A | p.A558E | |||
424743 | c.1683A>C | p.R561S | |||
7419559 | c.1748C>T | p.T583I | |||
239117 | c.1662A>C | p.E554D | |||
NM_001205293.1 | NP_001192222.1 | 6541384 | c.1682G>T | p.R561I | |
1133893 | c.1714C>T | p.R572* | |||
4569410 | c.1745T>A | p.I582K | |||
3376860 | c.1688G>T | p.G563V | |||
1583862 | c.1688G>A | p.G563D | |||
4668483 | c.1692G>A | p.T564T | |||
1199273 | c.1691C>T | p.T564M | |||
6541389 | c.1715G>T | p.R572L | |||
7419558 | c.1748C>T | p.T583I | |||
5527930 | c.1682G>A | p.R561K | |||
4452281 | c.1747A>C | p.T583P | |||
1133892 | c.1683A>C | p.R561S | |||
4925409 | c.1663G>A | p.V555M | |||
1134781 | c.1665G>T | p.V555V | |||
6894689 | c.1734A>C | p.R578S | |||
1336828 | c.1723C>T | p.R575W | |||
5784874 | c.1673C>A | p.A558E | |||
NM_001205294.1 | NP_001192223.1 | - | - | - | |
XM_017002243.1 | XP_016857732.1 | - | - | - | |
XM_017002244.1 | XP_016857733.1 | - | - | - | |
XM_017002245.1 | XP_016857734.1 | - | - | - | |
XM_017002246.1 | XP_016857735.1 | 7419560 | c.1601C>T | p.T534I | |
5160932 | c.1576C>T | p.R526W | |||
6894691 | c.1587A>C | p.R529S | |||
6541391 | c.1568G>T | p.R523L | |||
5784875 | c.1526C>A | p.A509E | |||
4925411 | c.1516G>A | p.V506M | |||
4668485 | c.1545G>A | p.T515T | |||
4569412 | c.1598T>A | p.I533K | |||
5527932 | c.1535G>A | p.R512K | |||
5214109 | c.1536A>C | p.R512S | |||
5208882 | c.1567C>T | p.R523* | |||
4452283 | c.1600A>C | p.T534P | |||
6541386 | c.1535G>T | p.R512I | |||
XM_017002247.1 | XP_016857736.1 | - | - | - | |
XM_017002248.1 | XP_016857737.1 | - | - | - | |
XM_017002249.1 | XP_016857738.1 | - | - | - | |
XM_017002250.1 | XP_016857739.1 | - | - | - | |
XM_017002251.1 | XP_016857740.1 | - | - | - | |
GenBank | AB209499.1 | BAD92736.1 | - | - | - |
ENST00000367570.0 | - | - | - | ||
L27745.1 | AAA72125.1 | - | - | - | |
L29384.1 | AAA59204.1 | - | - | - | |
L29385.1 | AAA59205.1 | - | - | - |
Literature Links: |
CACNA1E PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |