Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
LOC102724663 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NR_120491.1 | - | - | - | |
GenBank | BG186381.1 | - | - | - |
PPFIA2 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001220473.2 | NP_001207402.1 | - | - | - |
NM_001220474.2 | NP_001207403.1 | - | - | - | |
NM_001220475.2 | NP_001207404.1 | - | - | - | |
NM_001220476.2 | NP_001207405.1 | - | - | - | |
NM_001220477.2 | NP_001207406.1 | - | - | - | |
NM_001220478.2 | NP_001207407.1 | 3738567 | c.3011T>C | p.M1004T | |
4716872 | c.3091G>A | p.V1031I | |||
5923005 | c.3151G>A | p.V1051M | |||
4861922 | c.3166C>T | p.R1056* | |||
5498674 | c.3167+6T>C | p.? | |||
4487585 | c.3162C>T | p.F1054F | |||
5991932 | c.3126delA | p.D1043fs*6 | |||
6015082 | c.3154G>A | p.D1052N | |||
3465913 | c.2998G>A | p.A1000T | |||
6753317 | c.3049T>G | p.L1017V | |||
4780649 | c.3167+6T>G | p.? | |||
6881784 | c.3054G>A | p.G1018G | |||
5454177 | c.3098C>T | p.A1033V | |||
4861856 | c.3043C>G | p.P1015A | |||
3872599 | c.3023G>A | p.W1008* | |||
3738565 | c.3133C>T | p.R1045C | |||
7183490 | c.3029G>C | p.G1010A | |||
3710843 | c.3005G>T | p.G1002V | |||
6553555 | c.3064T>A | p.Y1022N | |||
4487354 | c.3132C>T | p.L1044L | |||
4866671 | c.3063G>T | p.Q1021H | |||
4211551 | c.3051G>T | p.L1017F | |||
4867276 | c.3101G>T | p.R1034I | |||
4857825 | c.3092T>C | p.V1031A | |||
7279998 | c.2998G>T | p.A1000S | |||
4867941 | c.3037T>A | p.W1013R | |||
6878586 | c.3029G>A | p.G1010E | |||
4869683 | c.3097G>A | p.A1033T | |||
4542227 | c.3101G>A | p.R1034K | |||
4045069 | c.3159T>C | p.S1053S | |||
3872597 | c.3034G>A | p.E1012K | |||
NM_001220479.2 | NP_001207408.1 | - | - | - | |
NM_001220480.2 | NP_001207409.1 | - | - | - | |
NM_001282536.1 | NP_001269465.1 | - | - | - | |
NM_003625.4 | NP_003616.2 | 5991931 | c.3171delA | p.D1058fs*6 | |
6015081 | c.3199G>A | p.D1067N | |||
3872598 | c.3068G>A | p.W1023* | |||
3710842 | c.3050G>T | p.G1017V | |||
287424 | c.3188T>G | p.L1063* | |||
695661 | c.3211C>T | p.R1071* | |||
468970 | c.3079G>A | p.E1027K | |||
943889 | c.3082T>A | p.W1028R | |||
395524 | c.3052G>A | p.D1018N | |||
1628875 | c.3212+6T>G | p.? | |||
3738564 | c.3178C>T | p.R1060C | |||
7183489 | c.3074G>C | p.G1025A | |||
943887 | c.3142G>A | p.A1048T | |||
4487584 | c.3207C>T | p.F1069F | |||
6878585 | c.3074G>A | p.G1025E | |||
3465912 | c.3043G>A | p.A1015T | |||
7279997 | c.3043G>T | p.A1015S | |||
1993553 | c.3096G>T | p.L1032F | |||
6553554 | c.3109T>A | p.Y1037N | |||
4045068 | c.3204T>C | p.S1068S | |||
4716871 | c.3136G>A | p.V1046I | |||
1221645 | c.3085C>T | p.L1029F | |||
6753316 | c.3094T>G | p.L1032V | |||
468969 | c.3137T>C | p.V1046A | |||
695660 | c.3088C>G | p.P1030A | |||
6881783 | c.3099G>A | p.G1033G | |||
943888 | c.3108G>T | p.Q1036H | |||
5498673 | c.3212+6T>C | p.? | |||
3738566 | c.3056T>C | p.M1019T | |||
943886 | c.3146G>T | p.R1049I | |||
4487353 | c.3177C>T | p.L1059L | |||
5454176 | c.3143C>T | p.A1048V | |||
4542226 | c.3146G>A | p.R1049K | |||
5923004 | c.3196G>A | p.V1066M | |||
XM_006719670.3 | XP_006719733.1 | - | - | - | |
XM_011538906.2 | XP_011537208.1 | - | - | - | |
XM_011538907.2 | XP_011537209.1 | - | - | - | |
XM_017020079.1 | XP_016875568.1 | - | - | - | |
XM_017020080.1 | XP_016875569.1 | - | - | - | |
XM_017020081.1 | XP_016875570.1 | - | - | - | |
XM_017020082.1 | XP_016875571.1 | - | - | - | |
XM_017020083.1 | XP_016875572.1 | - | - | - | |
XM_017020084.1 | XP_016875573.1 | - | - | - | |
XM_017020085.1 | XP_016875574.1 | - | - | - | |
XM_017020086.1 | XP_016875575.1 | - | - | - | |
XM_017020087.1 | XP_016875576.1 | - | - | - | |
XM_017020088.1 | XP_016875577.1 | - | - | - | |
XM_017020089.1 | XP_016875578.1 | - | - | - | |
XM_017020090.1 | XP_016875579.1 | - | - | - | |
XM_017020091.1 | XP_016875580.1 | - | - | - | |
XM_017020092.1 | XP_016875581.1 | - | - | - | |
XM_017020093.1 | XP_016875582.1 | - | - | - | |
XM_017020094.1 | XP_016875583.1 | - | - | - | |
XM_017020095.1 | XP_016875584.1 | - | - | - | |
XM_017020096.1 | XP_016875585.1 | - | - | - | |
XM_017020097.1 | XP_016875586.1 | - | - | - | |
XM_017020098.1 | XP_016875587.1 | - | - | - | |
XM_017020099.1 | XP_016875588.1 | - | - | - | |
XM_017020100.1 | XP_016875589.1 | - | - | - | |
XM_017020101.1 | XP_016875590.1 | - | - | - | |
XM_017020102.1 | XP_016875591.1 | - | - | - | |
XM_017020103.1 | XP_016875592.1 | - | - | - | |
XM_017020104.1 | XP_016875593.1 | - | - | - | |
XM_017020105.1 | XP_016875594.1 | - | - | - | |
XM_017020106.1 | XP_016875595.1 | - | - | - | |
XM_017020107.1 | XP_016875596.1 | - | - | - | |
XM_017020108.1 | XP_016875597.1 | - | - | - | |
XM_017020109.1 | XP_016875598.1 | - | - | - | |
XM_017020110.1 | XP_016875599.1 | - | - | - | |
XM_017020111.1 | XP_016875600.1 | - | - | - | |
XM_017020112.1 | XP_016875601.1 | - | - | - | |
XM_017020113.1 | XP_016875602.1 | - | - | - | |
XM_017020114.1 | XP_016875603.1 | - | - | - | |
XM_017020115.1 | XP_016875604.1 | - | - | - | |
XM_017020116.1 | XP_016875605.1 | - | - | - | |
XM_017020117.1 | XP_016875606.1 | - | - | - | |
XM_017020118.1 | XP_016875607.1 | - | - | - | |
XM_017020119.1 | XP_016875608.1 | - | - | - | |
XM_017020120.1 | XP_016875609.1 | - | - | - | |
GenBank | AB210009.1 | - | - | - | |
AF034799.1 | AAC26100.1 | - | - | - | |
AK123372.1 | - | - | - | ||
AK126971.1 | - | - | - | ||
AK294505.1 | - | - | - | ||
AK296380.1 | - | - | - | ||
AK297372.1 | - | - | - | ||
AK299853.1 | - | - | - | ||
AK302658.1 | - | - | - | ||
BC104912.1 | AAI04913.1 | - | - | - | |
BC143485.1 | - | - | - | ||
ENST00000549396.0 | - | - | - |
Literature Links: |
LOC102724663 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |