Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246674.2 | NP_001233603.1 | 1 - 2 | 83 | 59 | |
GenBank mRNA | JI866227.1 | - | 1 - 2 | 94 | 59 | |
JP060135.1 | - | 1 - 2 | 94 | 59 | ||
X57112.1 | - | 1 - 2 | 84 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |