Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159768.1 | NP_001153240.1 | 1 - 2 | 168 | 99 | |
NM_131163.2 | NP_571238.1 | 1 - 2 | 168 | 99 | ||
GenBank mRNA | BC062841.1 | - | 1 - 2 | 108 | 99 | |
BC162420.1 | - | 1 - 2 | 99 | 99 | ||
BC162435.1 | - | 1 - 2 | 99 | 99 | ||
L05383.1 | - | 1 - 2 | 109 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Endogenous Controls Recommended Assay Danio rerio Probe spans exons |