Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000590596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000590596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000590596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000590596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000590596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
ENST00000590596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007471.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002450.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002450.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002450.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002450.hg.1 |
TC18000213.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002446.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002446.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002446.hg.1 |
TC18000213.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002448.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002448.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002448.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002448.hg.1 |
TC18000213.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000590596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000590596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000590596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000590596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000590596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
ENST00000590596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007471.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800151267.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151267.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151267.hg.1 |
ENST00000590596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151267.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800151267.hg.1 |
TC1800007471.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800151267.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151267.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151267.hg.1 |
ENST00000590596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151267.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800151266.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151266.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151266.hg.1 |
ENST00000590596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151266.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800151266.hg.1 |
TC1800007471.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800151266.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151266.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151266.hg.1 |
ENST00000590596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151266.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800151271.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151271.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151271.hg.1 |
ENST00000590596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151271.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800151271.hg.1 |
TC1800007471.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800151271.hg.1 |
ENST00000269518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151271.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151271.hg.1 |
ENST00000590596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151271.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800151263.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151263.hg.1 |
NM_021127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800151263.hg.1 |
TC1800007471.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800151263.hg.1 |
ENST00000316660.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800151263.hg.1 |
NM_021127.2 |
Refseq |