Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110222.1 | NP_001103692.1 | 3 - 4 | 956 | 104 | |
NM_001110223.1 | NP_001103693.1 | 3 - 4 | 878 | 104 | ||
NM_001110224.1 | NP_001103694.1 | 3 - 4 | 878 | 104 | ||
NM_010025.2 | NP_034155.2 | 3 - 4 | 878 | 104 | ||
GenBank mRNA | AB011678.1 | - | 3 - 4 | 974 | 104 | |
AF045547.1 | - | 3 - 4 | 759 | 104 | ||
AK014264.1 | - | 3 - 4 | 879 | 104 | ||
AK048410.1 | - | 3 - 4 | 884 | 104 | ||
AK049050.1 | - | 3 - 4 | 952 | 104 | ||
AK164119.1 | - | 3 - 4 | 765 | 104 | ||
AY560329.1 | - | 3 - 4 | 766 | 104 | ||
BC056391.1 | - | 3 - 4 | 771 | 6855006 | 104 | |
BC057010.1 | - | 3 - 4 | 863 | 6825750 | 104 | |
BC062974.1 | - | 3 - 4 | 863 | 6825750 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |