Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nanog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289828.1 | NP_001276757.1 | 1 - 2 | 366 | 145 | |
NM_001289830.1 | NP_001276759.1 | 2 - 3 | 163 | 145 | ||
NM_001289831.1 | NP_001276760.1 | 2 - 3 | 147 | 145 | ||
NM_028016.3 | NP_082292.1 | 1 - 2 | 366 | 145 | ||
GenBank mRNA | AB093574.1 | - | 1 - 2 | 337 | 145 | |
AB126939.1 | - | 1 - 2 | 148 | 145 | ||
AF507043.1 | - | 1 - 2 | 338 | 145 | ||
AK010332.1 | - | 1 - 2 | 338 | 145 | ||
AK167259.1 | - | 1 - 2 | 337 | 145 | ||
AY278951.1 | - | 1 - 2 | 215 | 145 | ||
AY455282.1 | - | 2 - 3 | 147 | 145 | ||
AY455285.1 | - | 2 - 3 | 163 | 145 | ||
BC132017.1 | - | 1 - 2 | 363 | 40130294 | 145 | |
BC137873.1 | - | 1 - 2 | 363 | 8860892 | 145 | |
BC144995.1 | - | 1 - 2 | 363 | 9053543 | 145 | |
BC144996.1 | - | 1 - 2 | 363 | 9053544 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |