Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bdnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285422.1 | NP_001272351.1 | 1 | 455 | 77 | |
GenBank mRNA | EF125685.1 | - | 1 | 452 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |