Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256146.2 | XP_006256208.2 | 3 - 4 | 810 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |