Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107202.2 | NP_001100672.1 | 6 - 7 | 806 | 95 | |
XM_008769857.1 | XP_008768079.1 | 6 - 7 | 1096 | 95 | ||
GenBank mRNA | EU448292.1 | - | 6 - 7 | 806 | 95 | |
EU448293.1 | - | 6 - 7 | 806 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |