Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173940.2 | NP_776365.1 | 10 - 11 | 1337 | 88 | |
GenBank mRNA | AB060169.1 | - | 9 - 10 | 1713 | 88 | |
AF047692.1 | - | 9 - 10 | 1259 | 88 | ||
BC118109.1 | - | 10 - 11 | 1326 | 88 | ||
JQ766265.1 | - | 9 - 10 | 1239 | 88 | ||
U88329.1 | - | 10 - 11 | 1337 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |