Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174065.2 | NP_776490.2 | 5 - 6 | 898 | 81 | |
GenBank mRNA | BC119892.1 | - | 5 - 6 | 898 | 81 | |
Y08255.1 | - | 893 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |