Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629550.3 | XP_005629607.1 | 1685 | 75 | ||
GenBank mRNA | AY545218.2 | - | 1940 | 75 | ||
AY545219.2 | - | 2020 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris |