Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diablo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007628159.1 | XP_007626349.1 | 2 - 3 | 344 | 82 | |
GenBank mRNA | JI882455.1 | - | 2 - 3 | 344 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Probe spans exons |