Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641733.1 | XP_007639923.1 | 4 - 5 | 728 | 109 | |
GenBank mRNA | JI884472.1 | - | 4 - 5 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |