Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681761.3 | XP_005681818.3 | 3 - 4 | 690 | 74 | |
XM_013964723.2 | XP_013820177.2 | 3 - 4 | 911 | 74 | ||
XM_018049584.1 | XP_017905073.1 | 4 - 5 | 663 | 74 | ||
XM_018049585.1 | XP_017905074.1 | 4 - 5 | 663 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |