Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gapdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115114.1 | NP_001108586.1 | 11 - 12 | 993 | 100 | |
GenBank mRNA | AI106575.1 | - | 2 - 3 | 147 | 100 | |
BC083506.1 | - | 11 - 12 | 993 | 100 | ||
BC095386.1 | - | 11 - 12 | 993 | 100 | ||
BC115131.1 | - | 11 - 12 | 997 | 100 | ||
BI980143.1 | - | 1 - 2 | 316 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |