Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP3A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001329508.1 | NP_001316437.1 | 7 - 8 | 717 | 153 | |
XM_015294357.2 | XP_015149843.1 | 7 - 8 | 768 | 153 | ||
GenBank mRNA | BX931257.1 | - | 7 - 8 | 718 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |