Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589554.2 | XP_005589611.2 | 2 - 3 | 124 | 153 | |
GenBank mRNA | AK240626.1 | - | 2 - 3 | 112 | 153 | |
AK240631.1 | - | 2 - 3 | 110 | 153 | ||
X13887.1 | - | 125 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Probe spans exons |