Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172117.1 | NP_001165588.1 | 1 - 2 | 285 | 64 | |
NM_010407.4 | NP_034537.2 | 1 - 2 | 285 | 64 | ||
GenBank mRNA | AK149736.1 | - | 1 - 2 | 251 | 64 | |
AK150290.1 | - | 1 - 2 | 259 | 64 | ||
AK150709.1 | - | 1 - 2 | 268 | 64 | ||
AK152506.1 | - | 1 - 2 | 259 | 64 | ||
AK155975.1 | - | 1 - 2 | 198 | 64 | ||
AK159234.1 | - | 1 - 2 | 217 | 64 | ||
AK165315.1 | - | 1 - 2 | 250 | 64 | ||
BC010478.1 | - | 1 - 2 | 281 | 3978518 | 64 | |
J03023.1 | - | 1 - 2 | 174 | 64 | ||
Y00487.1 | - | 1 - 2 | 180 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |