Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diablo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023232.3 | NP_075721.3 | 2 - 3 | 217 | 85 | |
GenBank mRNA | AF203914.1 | - | 2 - 3 | 183 | 85 | |
AK002887.1 | - | 2 - 3 | 187 | 85 | ||
AK012760.1 | - | 2 - 3 | 215 | 85 | ||
AK077198.1 | - | 2 - 3 | 198 | 85 | ||
AK082903.1 | - | 2 - 3 | 187 | 85 | ||
AK133194.1 | - | 197 | 85 | |||
AK169573.1 | - | 2 - 3 | 187 | 85 | ||
BC024780.1 | - | 2 - 3 | 189 | 5365266 | 85 | |
BY080746.1 | - | 2 - 3 | 229 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |