Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010897.2 | NP_035027.1 | 29 - 30 | 4091 | 65 | |
GenBank mRNA | D30730.1 | - | 3 - 4 | 462 | 65 | |
D30731.1 | - | 3 - 4 | 462 | 65 | ||
L10370.1 | - | 28 - 29 | 3936 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |