Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013820.3 | NP_038848.1 | 9 - 10 | 1740 | 67 | |
GenBank mRNA | AJ238540.1 | - | 9 - 10 | 1683 | 67 | |
AK145936.1 | - | 9 - 10 | 1721 | 67 | ||
BC054472.1 | - | 9 - 10 | 1659 | 6465367 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |