Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009791.4 | NP_033921.3 | 24 - 25 | 8922 | 67 | |
GenBank mRNA | AF062378.1 | - | 7 - 8 | 1179 | 67 | |
AF533752.1 | - | 24 - 25 | 8900 | 67 | ||
AK048277.1 | - | 7 - 8 | 1735 | 67 | ||
AY971958.1 | - | 23 - 24 | 4968 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |