Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085364.4 | XP_001085364.1 | 2254 | 87 | ||
GenBank mRNA | JV479217.1 | - | 327 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta Inventoried |