Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053379.3 | NP_445831.3 | 3 - 4 | 772 | 103 | |
XM_006257382.2 | XP_006257444.1 | 3 - 4 | 897 | 103 | ||
XM_006257383.2 | XP_006257445.1 | 3 - 4 | 990 | 103 | ||
XM_006257384.2 | XP_006257446.1 | 3 - 4 | 989 | 103 | ||
XM_006257385.2 | XP_006257447.1 | 3 - 4 | 990 | 103 | ||
XM_006257386.2 | XP_006257448.1 | 3 - 4 | 990 | 103 | ||
XM_008773448.1 | XP_008771670.1 | 3 - 4 | 1459 | 103 | ||
GenBank mRNA | AF155959.2 | - | 2 - 3 | 706 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Inventoried Probe spans exons |