Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053528.1 | NP_445980.1 | 2 - 3 | 804 | 68 | |
XM_006229410.2 | XP_006229472.1 | 3 - 4 | 1145 | 68 | ||
XM_008759561.1 | XP_008757783.1 | 3 - 4 | 1147 | 68 | ||
XM_008759562.1 | XP_008757784.1 | 3 - 4 | 1091 | 68 | ||
GenBank mRNA | AJ245646.1 | - | 2 - 3 | 804 | 68 | |
AJ245647.1 | - | 2 - 3 | 804 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |