Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017045.1 | NP_058741.1 | 19 - 20 | 2252 | 97 | |
GenBank mRNA | D25233.1 | - | 19 - 20 | 2013 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |