Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss03395600_g1
See other GNAS GE Assays ›
Gene Symbol:
GNAS
Gene Name:
GNAS complex locus
Gene Aliases:
NESP55, PORGSA1, SCG6
Chromosome Location:
Chr.17: 58985580 - 59053022 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss03395600_g1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: GNAS
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001130215.2
NP_001123687.1
13
2265
140
NM_001171983.2
NP_001165454.1
12
1484
140
NM_001171984.2
NP_001165455.1
13
1529
140
NM_001171985.2
NP_001165456.1
12
1481
140
NM_214312.3
NP_999477.2
13
1526
140
NR_033291.2
13
1274
140
NR_033292.2
12
1229
140
NR_073434.1
12
1232
140
XM_021077150.1
XP_020932809.1
3595
140
XM_021077151.1
XP_020932810.1
3592
140
XM_021077152.1
XP_020932811.1
3550
140
XM_021077153.1
XP_020932812.1
3547
140
XM_021077154.1
XP_020932813.1
1174
140
XM_021077155.1
XP_020932814.1
2154
140
XM_021077156.1
XP_020932815.1
2217
140
XM_021077157.1
XP_020932816.1
1275
140
XM_021077158.1
XP_020932817.1
1234
140
XM_021077160.1
XP_020932819.1
2551
140
XM_021077161.1
XP_020932820.1
1020
140
XM_021077162.1
XP_020932821.1
1159
140
XM_021077163.1
XP_020932822.1
2506
140
XM_021077164.1
XP_020932823.1
1068
140
GenBank mRNA
AK233105.1
-
12
1176
140
AK237491.1
-
13
1229
140
AK238507.1
-
12
1187
140
AK238560.1
-
12
1187
140
AK238881.1
-
12
1128
140
AK240484.1
-
13
1260
140
AK347610.1
-
13
1110
140
AK348621.1
-
12
1176
140
AK390235.1
-
12
1146
140
AK392195.1
-
12
2154
140
AK394414.1
-
12
1098
140
AK396491.1
-
13
1270
140
AK398623.1
-
12
1186
140
AK400298.1
-
13
1229
140
AK400475.1
-
13
1233
140
AK400792.1
-
14
1416
140
X63893.1
-
140
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
3' Most
Detect Genomic DNA
Recommended Assay
Sus scrofa
Inventoried
Both primers and probe map within a single exon
Panther Classification:
Molecular Function -
heterotrimeric G-protein
Gene Ontology Categories:
Function(s)
Process(es)
adenylate cyclase-activating G-protein coupled receptor signaling pathway
positive regulation of GTPase activity
positive regulation of cAMP-mediated signaling
adenylate cyclase-activating adrenergic receptor signaling pathway
GTP binding
adenylate cyclase activator activity
metal ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-9jqhn:80/10.239.160.220:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0